Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41523172-41523397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO2
Entrez Gene ID
50
Gene Name
aconitase 2
Gene Aliases
ACONM, HEL-S-284, ICRD, OCA8, OPA9
UniGene
Hs.643610
Gene Chromosome Location
Chr.22: 41469125-41528989 on Build GRCh38
Transcripts
NM_001098.2 XM_017028812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1416527 c.1365G>A p.W455* Chr.22: 41523273-41523273 on Build GRCh38
1034638 c.1305C>A p.I435I Chr.22: 41523213-41523213 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764315098 Chr.22: 41523230-41523259 on Build GRCh38
rs376110432 Chr.22: 41523315-41523315 on Build GRCh38
rs980303518 Chr.22: 41523330-41523330 on Build GRCh38
rs985383080 Chr.22: 41523320-41523320 on Build GRCh38
rs927424471 Chr.22: 41523348-41523348 on Build GRCh38
rs748906180 Chr.22: 41523197-41523197 on Build GRCh38
rs374955715 Chr.22: 41523299-41523299 on Build GRCh38
rs1050922170 Chr.22: 41523261-41523261 on Build GRCh38
rs776637574 Chr.22: 41523288-41523288 on Build GRCh38
rs941475174 Chr.22: 41523350-41523350 on Build GRCh38
rs555720081 Chr.22: 41523354-41523354 on Build GRCh38
rs760581560 Chr.22: 41523246-41523246 on Build GRCh38
rs751717279 Chr.22: 41523288-41523289 on Build GRCh38
rs922920300 Chr.22: 41523356-41523356 on Build GRCh38
rs1042255265 Chr.22: 41523367-41523367 on Build GRCh38
rs773834741 Chr.22: 41523211-41523211 on Build GRCh38
rs145199596 Chr.22: 41523366-41523366 on Build GRCh38
rs775523355 Chr.22: 41523240-41523240 on Build GRCh38
rs763180569 Chr.22: 41523376-41523376 on Build GRCh38
rs770633599 Chr.22: 41523209-41523209 on Build GRCh38
rs12484694 Chr.22: 41523326-41523326 on Build GRCh38
rs763953506 Chr.22: 41523287-41523287 on Build GRCh38
rs918621442 Chr.22: 41523358-41523358 on Build GRCh38
rs371557782 Chr.22: 41523313-41523313 on Build GRCh38
rs745392648 Chr.22: 41523213-41523213 on Build GRCh38
rs771868586 Chr.22: 41523233-41523233 on Build GRCh38
rs1032662436 Chr.22: 41523220-41523220 on Build GRCh38
rs762114723 Chr.22: 41523296-41523296 on Build GRCh38

View Full Product Details