Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74712711-74712980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300LF
Entrez Gene ID
146722
Gene Name
CD300 molecule like family member f
Gene Aliases
CD300f, CLM-1, CLM1, IREM-1, IREM1, IgSF13, LMIR3, NKIR
UniGene
Hs.567706
Gene Chromosome Location
Chr.17: 74694308-74713059 on Build GRCh38
Transcripts
NM_001289082.1 NM_001289083.1 NM_001289084.1 NM_001289085.1 NM_001289086.1 NM_001289087.1 NM_139018.4 NR_110298.1 XM_017024212.1 XM_017024213.1 XM_017024214.1 XM_017024215.1 XM_017024216.1 XM_017024217.1 NM_175738.4

Gene Symbol
RAB37
Entrez Gene ID
326624
Gene Name
RAB37, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.351413
Gene Chromosome Location
Chr.17: 74671117-74747335 on Build GRCh38
Transcripts
NM_001289082.1 NM_001289083.1 NM_001289084.1 NM_001289085.1 NM_001289086.1 NM_001289087.1 NM_139018.4 NR_110298.1 XM_017024212.1 XM_017024213.1 XM_017024214.1 XM_017024215.1 XM_017024216.1 XM_017024217.1 NM_175738.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2977816 c.37C>T p.L13F Chr.17: 74712830-74712830 on Build GRCh38
5528365 c.4C>T p.P2S Chr.17: 74712863-74712863 on Build GRCh38
4069392 c.21C>A p.Y7* Chr.17: 74712846-74712846 on Build GRCh38
7414051 c.35G>T p.W12L Chr.17: 74712832-74712832 on Build GRCh38
5794138 c.21C>T p.Y7Y Chr.17: 74712846-74712846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs367985874 Chr.17: 74712812-74712812 on Build GRCh38
rs559558127 Chr.17: 74712760-74712760 on Build GRCh38
rs369800958 Chr.17: 74712884-74712884 on Build GRCh38
rs200814939 Chr.17: 74712888-74712888 on Build GRCh38
rs537461374 Chr.17: 74712937-74712937 on Build GRCh38
rs768088655 Chr.17: 74712760-74712761 on Build GRCh38
rs952605670 Chr.17: 74712845-74712845 on Build GRCh38
rs777038700 Chr.17: 74712887-74712887 on Build GRCh38
rs373467251 Chr.17: 74712745-74712745 on Build GRCh38
rs764895961 Chr.17: 74712786-74712786 on Build GRCh38
rs375445233 Chr.17: 74712865-74712865 on Build GRCh38
rs763753073 Chr.17: 74712924-74712924 on Build GRCh38
rs749927438 Chr.17: 74712798-74712798 on Build GRCh38
rs371126964 Chr.17: 74712904-74712904 on Build GRCh38
rs892029740 Chr.17: 74712733-74712733 on Build GRCh38
rs1004016149 Chr.17: 74712757-74712757 on Build GRCh38
rs748103558 Chr.17: 74712818-74712818 on Build GRCh38
rs750346658 Chr.17: 74712750-74712750 on Build GRCh38
rs751396558 Chr.17: 74712811-74712811 on Build GRCh38
rs772311900 Chr.17: 74712846-74712846 on Build GRCh38
rs199709522 Chr.17: 74712906-74712906 on Build GRCh38
rs747404024 Chr.17: 74712872-74712872 on Build GRCh38
rs1043214247 Chr.17: 74712747-74712747 on Build GRCh38
rs779808614 Chr.17: 74712810-74712810 on Build GRCh38
rs1043285161 Chr.17: 74712922-74712922 on Build GRCh38
rs1045059086 Chr.17: 74712746-74712746 on Build GRCh38
rs762363526 Chr.17: 74712923-74712923 on Build GRCh38
rs775631325 Chr.17: 74712861-74712861 on Build GRCh38
rs762507200 Chr.17: 74712913-74712913 on Build GRCh38
rs374495431 Chr.17: 74712808-74712808 on Build GRCh38
rs528248402 Chr.17: 74712771-74712771 on Build GRCh38
rs745918380 Chr.17: 74712833-74712833 on Build GRCh38
rs779014592 Chr.17: 74712830-74712830 on Build GRCh38
rs372170968 Chr.17: 74712817-74712817 on Build GRCh38
rs372574091 Chr.17: 74712885-74712885 on Build GRCh38
rs1035222285 Chr.17: 74712796-74712796 on Build GRCh38
rs946242435 Chr.17: 74712910-74712910 on Build GRCh38
rs758320992 Chr.17: 74712925-74712925 on Build GRCh38
rs904681143 Chr.17: 74712954-74712954 on Build GRCh38
rs769759652 Chr.17: 74712825-74712825 on Build GRCh38

View Full Product Details