Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73445066-73445299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFP
Entrez Gene ID
174
Gene Name
alpha fetoprotein
Gene Aliases
AFPD, FETA, HPAFP
UniGene
Hs.518808
Gene Chromosome Location
Chr.4: 73436219-73455785 on Build GRCh38
Transcripts
NM_001134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3409471 c.793G>T p.V265L Chr.4: 73445072-73445072 on Build GRCh38
7245815 c.843+5A>G p.? Chr.4: 73445127-73445127 on Build GRCh38
5401512 c.838G>A p.D280N Chr.4: 73445117-73445117 on Build GRCh38
6266375 c.841G>T p.G281W Chr.4: 73445120-73445120 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772835409 Chr.4: 73445141-73445141 on Build GRCh38
rs538380590 Chr.4: 73445246-73445246 on Build GRCh38
rs938481978 Chr.4: 73445179-73445179 on Build GRCh38
rs868825016 Chr.4: 73445228-73445228 on Build GRCh38
rs752855382 Chr.4: 73445155-73445156 on Build GRCh38
rs777404284 Chr.4: 73445160-73445161 on Build GRCh38
rs373601020 Chr.4: 73445130-73445130 on Build GRCh38
rs914253982 Chr.4: 73445129-73445129 on Build GRCh38
rs759634221 Chr.4: 73445147-73445149 on Build GRCh38
rs755991068 Chr.4: 73445158-73445159 on Build GRCh38
rs780261526 Chr.4: 73445103-73445103 on Build GRCh38
rs1056035614 Chr.4: 73445192-73445192 on Build GRCh38
rs79284391 Chr.4: 73445200-73445200 on Build GRCh38
rs771478094 Chr.4: 73445135-73445135 on Build GRCh38
rs1037693746 Chr.4: 73445214-73445214 on Build GRCh38
rs746897337 Chr.4: 73445152-73445152 on Build GRCh38
rs928470987 Chr.4: 73445146-73445146 on Build GRCh38
rs1030622463 Chr.4: 73445242-73445242 on Build GRCh38
rs544812935 Chr.4: 73445215-73445215 on Build GRCh38
rs767859286 Chr.4: 73445154-73445155 on Build GRCh38
rs770939080 Chr.4: 73445153-73445153 on Build GRCh38
rs572667172 Chr.4: 73445189-73445189 on Build GRCh38
rs894535023 Chr.4: 73445199-73445199 on Build GRCh38
rs571205594 Chr.4: 73445221-73445221 on Build GRCh38
rs749712578 Chr.4: 73445120-73445120 on Build GRCh38
rs776578845 Chr.4: 73445163-73445163 on Build GRCh38
rs753850065 Chr.4: 73445161-73445162 on Build GRCh38
rs750893628 Chr.4: 73445091-73445091 on Build GRCh38
rs993748023 Chr.4: 73445237-73445237 on Build GRCh38
rs755427022 Chr.4: 73445125-73445125 on Build GRCh38
rs756705227 Chr.4: 73445099-73445099 on Build GRCh38
rs997269440 Chr.4: 73445178-73445178 on Build GRCh38
rs2298839 Chr.4: 73445127-73445127 on Build GRCh38
rs371069157 Chr.4: 73445144-73445144 on Build GRCh38
rs954949771 Chr.4: 73445266-73445266 on Build GRCh38

View Full Product Details