Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219514958-219515231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC4
Entrez Gene ID
55515
Gene Name
acid sensing ion channel subunit family member 4
Gene Aliases
ACCN4, BNAC4
UniGene
Hs.87469
Gene Chromosome Location
Chr.2: 219514170-219538772 on Build GRCh38
Transcripts
NM_182847.2 XM_005246671.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4937024 c.758C>A p.S253* Chr.2: 219515101-219515101 on Build GRCh38
7292431 c.803G>T p.G268V Chr.2: 219515146-219515146 on Build GRCh38
7319819 c.848G>A p.R283H Chr.2: 219515191-219515191 on Build GRCh38
7323679 c.879C>G p.L293L Chr.2: 219515222-219515222 on Build GRCh38
7356602 c.646G>T p.A216S Chr.2: 219514989-219514989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780645754 Chr.2: 219515050-219515050 on Build GRCh38
rs779639143 Chr.2: 219515127-219515127 on Build GRCh38
rs140032110 Chr.2: 219515117-219515117 on Build GRCh38
rs749067696 Chr.2: 219515131-219515131 on Build GRCh38
rs765419366 Chr.2: 219515091-219515091 on Build GRCh38
rs36089490 Chr.2: 219515105-219515105 on Build GRCh38
rs767948018 Chr.2: 219515200-219515200 on Build GRCh38
rs754102861 Chr.2: 219515029-219515029 on Build GRCh38
rs746558898 Chr.2: 219514979-219514979 on Build GRCh38
rs967035531 Chr.2: 219515190-219515190 on Build GRCh38
rs755637023 Chr.2: 219515211-219515211 on Build GRCh38
rs749947411 Chr.2: 219515118-219515118 on Build GRCh38
rs778494559 Chr.2: 219515039-219515039 on Build GRCh38
rs1035507956 Chr.2: 219515037-219515037 on Build GRCh38
rs753103048 Chr.2: 219515095-219515095 on Build GRCh38
rs575903196 Chr.2: 219514992-219514992 on Build GRCh38
rs776669347 Chr.2: 219515081-219515081 on Build GRCh38
rs746507588 Chr.2: 219515047-219515047 on Build GRCh38
rs762968010 Chr.2: 219515008-219515008 on Build GRCh38
rs745596053 Chr.2: 219515004-219515004 on Build GRCh38
rs142235643 Chr.2: 219515052-219515052 on Build GRCh38
rs760689772 Chr.2: 219515072-219515072 on Build GRCh38
rs771952448 Chr.2: 219515175-219515175 on Build GRCh38
rs770046746 Chr.2: 219515179-219515179 on Build GRCh38
rs368212523 Chr.2: 219515003-219515003 on Build GRCh38
rs146293263 Chr.2: 219515069-219515069 on Build GRCh38
rs146925695 Chr.2: 219515033-219515033 on Build GRCh38
rs752059801 Chr.2: 219515192-219515192 on Build GRCh38
rs764587492 Chr.2: 219515191-219515191 on Build GRCh38
rs748107085 Chr.2: 219515163-219515163 on Build GRCh38
rs1049761394 Chr.2: 219515137-219515137 on Build GRCh38
rs760809171 Chr.2: 219515023-219515023 on Build GRCh38
rs766574644 Chr.2: 219515025-219515025 on Build GRCh38
rs749942049 Chr.2: 219515201-219515201 on Build GRCh38
rs759781609 Chr.2: 219515086-219515086 on Build GRCh38
rs759602562 Chr.2: 219515176-219515176 on Build GRCh38
rs115934162 Chr.2: 219515034-219515034 on Build GRCh38
rs35780339 Chr.2: 219515063-219515063 on Build GRCh38
rs758944408 Chr.2: 219515038-219515038 on Build GRCh38
rs749118047 Chr.2: 219515062-219515062 on Build GRCh38
rs1032499548 Chr.2: 219515104-219515104 on Build GRCh38
rs762384202 Chr.2: 219515196-219515196 on Build GRCh38
rs764640541 Chr.2: 219515102-219515102 on Build GRCh38
rs764065064 Chr.2: 219515013-219515013 on Build GRCh38
rs945783346 Chr.2: 219514980-219514980 on Build GRCh38
rs147937668 Chr.2: 219515132-219515132 on Build GRCh38
rs147551339 Chr.2: 219515183-219515183 on Build GRCh38
rs777402491 Chr.2: 219514978-219514978 on Build GRCh38
rs975816858 Chr.2: 219514993-219514993 on Build GRCh38
rs745815024 Chr.2: 219515161-219515172 on Build GRCh38
rs941961367 Chr.2: 219515030-219515030 on Build GRCh38
rs766449710 Chr.2: 219515079-219515079 on Build GRCh38
rs773243756 Chr.2: 219515015-219515015 on Build GRCh38
rs770706430 Chr.2: 219514995-219514995 on Build GRCh38
rs755757405 Chr.2: 219515122-219515122 on Build GRCh38
rs756791914 Chr.2: 219515114-219515114 on Build GRCh38
rs769639891 Chr.2: 219515006-219515006 on Build GRCh38
rs138559599 Chr.2: 219515053-219515053 on Build GRCh38
rs758893039 Chr.2: 219515101-219515101 on Build GRCh38
rs202143794 Chr.2: 219515007-219515007 on Build GRCh38
rs868697709 Chr.2: 219515133-219515133 on Build GRCh38
rs775871563 Chr.2: 219515180-219515180 on Build GRCh38
rs756845255 Chr.2: 219515049-219515049 on Build GRCh38

View Full Product Details