Product Details
- Primer Sequences
- Log in to view
- Primer Chromosome Location
-
Chr.1:
145728505-145728238
on Build GRCh38
View in UCSC browser
- Format
- Each primer will be delivered in a single tube, dried-down
- Quick Reference Guide
- Primer Designer™ Tool Quick Reference
- Ampliseq ID's
Gene Details
- Gene Symbol
- CD160
- Entrez Gene ID
-
11126
- Gene Name
- CD160 molecule
- Gene Aliases
-
BY55, NK1, NK28
- UniGene
- Hs.488237
- Gene Chromosome Location
-
Chr.1:
145719312-145739288
on Build GRCh38
- Transcripts
-
NM_007053.3
NR_103845.1
XM_005272929.3
XM_011509104.2
Cosmic Detail
Cosmic ID |
Mutation |
AA |
Transcript Location |
5349952 |
c.16G>A |
p.G6S |
Chr.1:
145728343-145728343
on Build GRCh38
|
674776 |
c.71G>C |
p.G24A |
Chr.1:
145728398-145728398
on Build GRCh38
|
5587456 |
c.9G>A |
p.L3L |
Chr.1:
145728336-145728336
on Build GRCh38
|
5577318 |
c.20G>A |
p.R7K |
Chr.1:
145728347-145728347
on Build GRCh38
|
View Full Product Details