Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 40180782-40181043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUGCT
Entrez Gene ID
79783
Gene Name
succinyl-CoA:glutarate-CoA transferase
Gene Aliases
C7orf10, DERP13, GA3, ORF19
UniGene
Hs.586313
Gene Chromosome Location
Chr.7: 40134887-40860767 on Build GRCh38
Transcripts
NM_001193311.1 NM_001193312.1 NM_001193313.1 NM_024728.2 XM_006715775.3 XM_011515525.2 XM_011515526.2 XM_011515527.2 XM_011515528.2 XM_011515529.2 XM_011515530.2 XM_017012621.1 XM_017012622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4826970 c.160C>G p.L54V Chr.7: 40180985-40180985 on Build GRCh38
5079457 c.133A>C p.K45Q Chr.7: 40180979-40180979 on Build GRCh38
3638639 c.134T>C p.I45T Chr.7: 40180959-40180959 on Build GRCh38
5216146 c.101-1G>C p.? Chr.7: 40180946-40180946 on Build GRCh38
3638641 c.167T>G p.L56R Chr.7: 40180992-40180992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116844030 Chr.7: 40180876-40180876 on Build GRCh38
rs778031775 Chr.7: 40181005-40181005 on Build GRCh38
rs368555179 Chr.7: 40180980-40180980 on Build GRCh38
rs779607926 Chr.7: 40180900-40180900 on Build GRCh38
rs199787147 Chr.7: 40180920-40180920 on Build GRCh38
rs969188163 Chr.7: 40181004-40181004 on Build GRCh38
rs925664844 Chr.7: 40180847-40180847 on Build GRCh38
rs753255609 Chr.7: 40180901-40180901 on Build GRCh38
rs755650226 Chr.7: 40180897-40180897 on Build GRCh38
rs372464576 Chr.7: 40180949-40180949 on Build GRCh38
rs757540185 Chr.7: 40181009-40181009 on Build GRCh38
rs368613630 Chr.7: 40180922-40180922 on Build GRCh38
rs774144146 Chr.7: 40180968-40180968 on Build GRCh38
rs775471728 Chr.7: 40180956-40180956 on Build GRCh38
rs375965592 Chr.7: 40181002-40181002 on Build GRCh38
rs73687513 Chr.7: 40180896-40180896 on Build GRCh38
rs751997717 Chr.7: 40181006-40181006 on Build GRCh38
rs7778061 Chr.7: 40180813-40180813 on Build GRCh38
rs561178672 Chr.7: 40180967-40180967 on Build GRCh38
rs201425078 Chr.7: 40180999-40180999 on Build GRCh38
rs781241078 Chr.7: 40180935-40180935 on Build GRCh38
rs553056400 Chr.7: 40180836-40180836 on Build GRCh38
rs193023834 Chr.7: 40180972-40180972 on Build GRCh38
rs913364377 Chr.7: 40180846-40180846 on Build GRCh38
rs543382689 Chr.7: 40180996-40180996 on Build GRCh38
rs374236126 Chr.7: 40180913-40180913 on Build GRCh38
rs746009032 Chr.7: 40180944-40180944 on Build GRCh38
rs762809682 Chr.7: 40180963-40180963 on Build GRCh38
rs577980239 Chr.7: 40180988-40180988 on Build GRCh38
rs1013453163 Chr.7: 40180992-40180992 on Build GRCh38
rs557100731 Chr.7: 40181003-40181003 on Build GRCh38
rs768451436 Chr.7: 40180964-40180964 on Build GRCh38
rs2036041 Chr.7: 40180908-40180908 on Build GRCh38
rs1040850842 Chr.7: 40180886-40180886 on Build GRCh38

View Full Product Details