Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95956652-95956893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMDHD1
Entrez Gene ID
144193
Gene Name
amidohydrolase domain containing 1
Gene Aliases
HMFT1272
UniGene
Hs.424907
Gene Chromosome Location
Chr.12: 95943293-95968592 on Build GRCh38
Transcripts
NM_152435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2046459 c.452C>T p.T151M Chr.12: 95956827-95956827 on Build GRCh38
7236503 c.389C>T p.T130I Chr.12: 95956764-95956764 on Build GRCh38
7236648 c.426C>G p.L142L Chr.12: 95956801-95956801 on Build GRCh38
265990 c.458T>C p.V153A Chr.12: 95956833-95956833 on Build GRCh38
6138220 c.421C>A p.R141R Chr.12: 95956796-95956796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755703276 Chr.12: 95956747-95956747 on Build GRCh38
rs746297553 Chr.12: 95956757-95956757 on Build GRCh38
rs753514490 Chr.12: 95956876-95956876 on Build GRCh38
rs779552415 Chr.12: 95956755-95956755 on Build GRCh38
rs145626384 Chr.12: 95956715-95956715 on Build GRCh38
rs530496824 Chr.12: 95956742-95956742 on Build GRCh38
rs764118926 Chr.12: 95956704-95956704 on Build GRCh38
rs551304292 Chr.12: 95956779-95956779 on Build GRCh38
rs751426964 Chr.12: 95956727-95956727 on Build GRCh38
rs779518192 Chr.12: 95956815-95956815 on Build GRCh38
rs766353218 Chr.12: 95956781-95956781 on Build GRCh38
rs772293939 Chr.12: 95956760-95956760 on Build GRCh38
rs758813223 Chr.12: 95956832-95956832 on Build GRCh38
rs748266716 Chr.12: 95956851-95956851 on Build GRCh38
rs201374830 Chr.12: 95956812-95956812 on Build GRCh38
rs759403725 Chr.12: 95956787-95956787 on Build GRCh38
rs763890532 Chr.12: 95956871-95956871 on Build GRCh38
rs369034222 Chr.12: 95956780-95956780 on Build GRCh38
rs766298210 Chr.12: 95956723-95956723 on Build GRCh38
rs567215522 Chr.12: 95956870-95956870 on Build GRCh38
rs767346412 Chr.12: 95956865-95956865 on Build GRCh38
rs925143393 Chr.12: 95956795-95956795 on Build GRCh38
rs746321378 Chr.12: 95956827-95956827 on Build GRCh38
rs752248228 Chr.12: 95956741-95956741 on Build GRCh38
rs749701037 Chr.12: 95956842-95956842 on Build GRCh38
rs534815977 Chr.12: 95956849-95956849 on Build GRCh38
rs767164703 Chr.12: 95956790-95956790 on Build GRCh38
rs763445534 Chr.12: 95956749-95956749 on Build GRCh38
rs375853196 Chr.12: 95956703-95956703 on Build GRCh38
rs201085031 Chr.12: 95956749-95956749 on Build GRCh38
rs527424065 Chr.12: 95956818-95956818 on Build GRCh38
rs768922385 Chr.12: 95956844-95956844 on Build GRCh38
rs754706271 Chr.12: 95956733-95956733 on Build GRCh38
rs762821619 Chr.12: 95956720-95956720 on Build GRCh38
rs775813472 Chr.12: 95956773-95956773 on Build GRCh38
rs757909459 Chr.12: 95956754-95956754 on Build GRCh38
rs774124190 Chr.12: 95956783-95956783 on Build GRCh38
rs772400510 Chr.12: 95956690-95956690 on Build GRCh38
rs769824213 Chr.12: 95956854-95956854 on Build GRCh38
rs200378404 Chr.12: 95956806-95956806 on Build GRCh38
rs373316462 Chr.12: 95956861-95956861 on Build GRCh38
rs200706449 Chr.12: 95956859-95956859 on Build GRCh38
rs775212542 Chr.12: 95956868-95956868 on Build GRCh38
rs1055064885 Chr.12: 95956802-95956802 on Build GRCh38
rs773837719 Chr.12: 95956674-95956675 on Build GRCh38
rs759816920 Chr.12: 95956679-95956679 on Build GRCh38
rs780936530 Chr.12: 95956738-95956738 on Build GRCh38
rs982843448 Chr.12: 95956730-95956730 on Build GRCh38
rs151046034 Chr.12: 95956793-95956793 on Build GRCh38
rs112657735 Chr.12: 95956837-95956837 on Build GRCh38
rs199693405 Chr.12: 95956808-95956808 on Build GRCh38
rs755758291 Chr.12: 95956801-95956801 on Build GRCh38
rs143138689 Chr.12: 95956834-95956834 on Build GRCh38
rs928634093 Chr.12: 95956748-95956748 on Build GRCh38
rs966486330 Chr.12: 95956792-95956792 on Build GRCh38
rs771138674 Chr.12: 95956776-95956777 on Build GRCh38

View Full Product Details