Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38126491-38126744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAA1
Entrez Gene ID
30
Gene Name
acetyl-CoA acyltransferase 1
Gene Aliases
ACAA, PTHIO, THIO
UniGene
Hs.643487
Gene Chromosome Location
Chr.3: 38122710-38137242 on Build GRCh38
Transcripts
NM_001130410.1 NM_001607.3 NR_024024.1 XM_006713122.1 XM_006713123.2 XM_011533650.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5864419 c.296G>A p.S99N Chr.3: 38126575-38126575 on Build GRCh38
5864418 c.752G>A p.S251N Chr.3: 38126575-38126575 on Build GRCh38
6276533 c.251C>A p.T84N Chr.3: 38126620-38126620 on Build GRCh38
7194811 c.800A>G p.D267G Chr.3: 38126527-38126527 on Build GRCh38
1044147 c.746G>A p.R249H Chr.3: 38126581-38126581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761579086 Chr.3: 38126559-38126559 on Build GRCh38
rs150485297 Chr.3: 38126535-38126535 on Build GRCh38
rs970031657 Chr.3: 38126698-38126698 on Build GRCh38
rs755386041 Chr.3: 38126587-38126587 on Build GRCh38
rs778020956 Chr.3: 38126628-38126628 on Build GRCh38
rs773054135 Chr.3: 38126699-38126699 on Build GRCh38
rs555852609 Chr.3: 38126658-38126658 on Build GRCh38
rs370502860 Chr.3: 38126519-38126519 on Build GRCh38
rs761738586 Chr.3: 38126670-38126670 on Build GRCh38
rs115896366 Chr.3: 38126581-38126581 on Build GRCh38
rs750066523 Chr.3: 38126569-38126569 on Build GRCh38
rs140435043 Chr.3: 38126582-38126582 on Build GRCh38
rs778391520 Chr.3: 38126656-38126656 on Build GRCh38
rs763063391 Chr.3: 38126514-38126514 on Build GRCh38
rs200810325 Chr.3: 38126713-38126713 on Build GRCh38
rs765930950 Chr.3: 38126719-38126719 on Build GRCh38
rs933541715 Chr.3: 38126544-38126544 on Build GRCh38
rs113334896 Chr.3: 38126623-38126623 on Build GRCh38
rs771110680 Chr.3: 38126640-38126640 on Build GRCh38
rs376130915 Chr.3: 38126659-38126659 on Build GRCh38
rs778312939 Chr.3: 38126558-38126568 on Build GRCh38
rs752171275 Chr.3: 38126583-38126583 on Build GRCh38
rs919532888 Chr.3: 38126539-38126539 on Build GRCh38
rs760526334 Chr.3: 38126538-38126538 on Build GRCh38
rs149277599 Chr.3: 38126589-38126589 on Build GRCh38
rs774228565 Chr.3: 38126641-38126641 on Build GRCh38
rs917285306 Chr.3: 38126682-38126682 on Build GRCh38
rs992938366 Chr.3: 38126667-38126667 on Build GRCh38
rs776597134 Chr.3: 38126661-38126661 on Build GRCh38
rs910547843 Chr.3: 38126564-38126564 on Build GRCh38
rs202236274 Chr.3: 38126681-38126681 on Build GRCh38
rs750987835 Chr.3: 38126721-38126721 on Build GRCh38
rs763955025 Chr.3: 38126558-38126558 on Build GRCh38
rs61095322 Chr.3: 38126634-38126634 on Build GRCh38
rs765100298 Chr.3: 38126685-38126685 on Build GRCh38
rs748475139 Chr.3: 38126602-38126602 on Build GRCh38
rs2011003 Chr.3: 38126562-38126562 on Build GRCh38

View Full Product Details