Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48799778-48800023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAT2
Entrez Gene ID
587
Gene Name
branched chain amino acid transaminase 2
Gene Aliases
BCAM, BCATM, BCT2, PP18
UniGene
Hs.512670
Gene Chromosome Location
Chr.19: 48795062-48811063 on Build GRCh38
Transcripts
NM_001164773.1 NM_001190.3 NM_001284325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3757018 c.557C>G p.T186R Chr.19: 48799813-48799813 on Build GRCh38
1283641 c.536C>T p.S179L Chr.19: 48799834-48799834 on Build GRCh38
999085 c.490G>A p.G164S Chr.19: 48800022-48800022 on Build GRCh38
6568158 c.528C>T p.N176N Chr.19: 48799984-48799984 on Build GRCh38
3823565 c.493A>C p.T165P Chr.19: 48800019-48800019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768593940 Chr.19: 48799808-48799808 on Build GRCh38
rs753566808 Chr.19: 48799946-48799946 on Build GRCh38
rs138273695 Chr.19: 48799923-48799924 on Build GRCh38
rs1033304218 Chr.19: 48799913-48799913 on Build GRCh38
rs201078371 Chr.19: 48799977-48799977 on Build GRCh38
rs79924510 Chr.19: 48799881-48799881 on Build GRCh38
rs1045285691 Chr.19: 48799838-48799838 on Build GRCh38
rs751627246 Chr.19: 48799834-48799834 on Build GRCh38
rs749866079 Chr.19: 48800003-48800003 on Build GRCh38
rs749674051 Chr.19: 48799845-48799845 on Build GRCh38
rs1022339040 Chr.19: 48799968-48799968 on Build GRCh38
rs773653450 Chr.19: 48799840-48799840 on Build GRCh38
rs757000896 Chr.19: 48799947-48799947 on Build GRCh38
rs1010922800 Chr.19: 48799967-48799967 on Build GRCh38
rs781079074 Chr.19: 48799976-48799976 on Build GRCh38
rs778419323 Chr.19: 48799993-48799993 on Build GRCh38
rs911136797 Chr.19: 48799999-48799999 on Build GRCh38
rs377427799 Chr.19: 48799861-48799861 on Build GRCh38
rs1042293172 Chr.19: 48799879-48799879 on Build GRCh38
rs894905096 Chr.19: 48799943-48799943 on Build GRCh38
rs771392617 Chr.19: 48799853-48799853 on Build GRCh38
rs756622271 Chr.19: 48799988-48799988 on Build GRCh38
rs193099355 Chr.19: 48799983-48799983 on Build GRCh38
rs373806842 Chr.19: 48799882-48799882 on Build GRCh38
rs748621319 Chr.19: 48799814-48799814 on Build GRCh38
rs773860470 Chr.19: 48799859-48799859 on Build GRCh38
rs765140805 Chr.19: 48799948-48799948 on Build GRCh38
rs1048183673 Chr.19: 48799809-48799809 on Build GRCh38
rs1025165159 Chr.19: 48799966-48799966 on Build GRCh38
rs751697837 Chr.19: 48799955-48799955 on Build GRCh38
rs758664912 Chr.19: 48799950-48799950 on Build GRCh38
rs533276867 Chr.19: 48799877-48799877 on Build GRCh38
rs11548193 Chr.19: 48799813-48799813 on Build GRCh38
rs755257830 Chr.19: 48799957-48799957 on Build GRCh38
rs986571893 Chr.19: 48799984-48799984 on Build GRCh38
rs766907460 Chr.19: 48799868-48799868 on Build GRCh38
rs747326261 Chr.19: 48799806-48799806 on Build GRCh38
rs1012327340 Chr.19: 48799822-48799822 on Build GRCh38
rs771179659 Chr.19: 48800006-48800006 on Build GRCh38

View Full Product Details