Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32391926-32392199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
UniGene
Hs.150107
Gene Chromosome Location
Chr.2: 32356907-32618899 on Build GRCh38
Transcripts
NM_016252.3 XM_005264449.4 XM_005264450.4 XM_005264451.4 XM_005264452.4 XM_005264453.4 XM_005264454.4 XM_005264455.4 XM_006712054.3 XM_006712055.3 XM_006712056.3 XM_011533003.2 XM_011533005.2 XM_017004556.1 XM_017004557.1 XM_017004558.1 XM_017004559.1 XM_017004560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5646024 c.884C>G p.S295* Chr.2: 32392083-32392083 on Build GRCh38
6841249 c.868C>T p.R290W Chr.2: 32392067-32392067 on Build GRCh38
69821 c.905G>C p.R302T Chr.2: 32392104-32392104 on Build GRCh38
7208353 c.908G>A p.W303* Chr.2: 32392107-32392107 on Build GRCh38
5396930 c.934G>A p.A312T Chr.2: 32392133-32392133 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757046184 Chr.2: 32392017-32392017 on Build GRCh38
rs535861090 Chr.2: 32391978-32391978 on Build GRCh38
rs1030639573 Chr.2: 32392060-32392060 on Build GRCh38
rs767772976 Chr.2: 32391990-32391990 on Build GRCh38
rs908832741 Chr.2: 32391979-32391979 on Build GRCh38
rs758332196 Chr.2: 32392059-32392059 on Build GRCh38
rs866592973 Chr.2: 32392116-32392116 on Build GRCh38
rs982211337 Chr.2: 32392002-32392002 on Build GRCh38
rs779906155 Chr.2: 32392067-32392067 on Build GRCh38
rs754470352 Chr.2: 32392097-32392097 on Build GRCh38
rs771257401 Chr.2: 32392129-32392129 on Build GRCh38
rs566826081 Chr.2: 32391968-32391968 on Build GRCh38
rs1026964908 Chr.2: 32391950-32391950 on Build GRCh38
rs576506192 Chr.2: 32391961-32391961 on Build GRCh38
rs773898677 Chr.2: 32392040-32392040 on Build GRCh38
rs774853197 Chr.2: 32392160-32392160 on Build GRCh38
rs374496224 Chr.2: 32392165-32392165 on Build GRCh38
rs746169660 Chr.2: 32392161-32392161 on Build GRCh38
rs764998594 Chr.2: 32392031-32392031 on Build GRCh38
rs78216644 Chr.2: 32392006-32392006 on Build GRCh38
rs376677624 Chr.2: 32392008-32392008 on Build GRCh38
rs1055480813 Chr.2: 32392015-32392015 on Build GRCh38
rs62136276 Chr.2: 32391984-32391984 on Build GRCh38
rs914207595 Chr.2: 32392018-32392018 on Build GRCh38
rs746541448 Chr.2: 32392068-32392068 on Build GRCh38
rs747813818 Chr.2: 32392123-32392123 on Build GRCh38
rs956681648 Chr.2: 32392155-32392155 on Build GRCh38
rs111335873 Chr.2: 32392013-32392013 on Build GRCh38
rs772720622 Chr.2: 32392174-32392174 on Build GRCh38
rs780745775 Chr.2: 32392117-32392117 on Build GRCh38

View Full Product Details