Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35871935-35872162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APLP1
Entrez Gene ID
333
Gene Name
amyloid beta precursor like protein 1
Gene Aliases
APLP
UniGene
Hs.74565
Gene Chromosome Location
Chr.19: 35868445-35879797 on Build GRCh38
Transcripts
NM_001024807.2 NM_005166.4 XM_017026737.1 XM_017026738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
995305 c.829A>G p.T277A Chr.19: 35872015-35872015 on Build GRCh38
6602199 c.758T>C p.V253A Chr.19: 35871944-35871944 on Build GRCh38
159123 c.826C>T p.H276Y Chr.19: 35872012-35872012 on Build GRCh38
995303 c.771C>T p.F257F Chr.19: 35871957-35871957 on Build GRCh38
995304 c.782C>T p.P261L Chr.19: 35871968-35871968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015003710 Chr.19: 35872055-35872055 on Build GRCh38
rs777128414 Chr.19: 35872018-35872019 on Build GRCh38
rs1014319518 Chr.19: 35872054-35872054 on Build GRCh38
rs1016965301 Chr.19: 35872133-35872133 on Build GRCh38
rs143738110 Chr.19: 35872024-35872024 on Build GRCh38
rs973398345 Chr.19: 35872072-35872072 on Build GRCh38
rs892180063 Chr.19: 35871994-35871994 on Build GRCh38
rs369846598 Chr.19: 35871969-35871969 on Build GRCh38
rs900038509 Chr.19: 35872071-35872071 on Build GRCh38
rs764892199 Chr.19: 35872026-35872026 on Build GRCh38
rs766578121 Chr.19: 35872014-35872014 on Build GRCh38
rs1043674357 Chr.19: 35871992-35871992 on Build GRCh38
rs963023252 Chr.19: 35872136-35872136 on Build GRCh38
rs376386528 Chr.19: 35872047-35872047 on Build GRCh38
rs369421258 Chr.19: 35872006-35872006 on Build GRCh38
rs777735772 Chr.19: 35871968-35871968 on Build GRCh38
rs1029764244 Chr.19: 35872089-35872089 on Build GRCh38
rs780812000 Chr.19: 35871987-35871987 on Build GRCh38
rs199883121 Chr.19: 35872030-35872030 on Build GRCh38
rs1018934881 Chr.19: 35872060-35872060 on Build GRCh38
rs982846182 Chr.19: 35872110-35872110 on Build GRCh38
rs752514509 Chr.19: 35872027-35872027 on Build GRCh38
rs760822143 Chr.19: 35872009-35872009 on Build GRCh38
rs372531049 Chr.19: 35872000-35872000 on Build GRCh38
rs540130957 Chr.19: 35872079-35872079 on Build GRCh38
rs953721382 Chr.19: 35872117-35872117 on Build GRCh38
rs112608328 Chr.19: 35871996-35871996 on Build GRCh38
rs753886590 Chr.19: 35872029-35872029 on Build GRCh38
rs756829664 Chr.19: 35871982-35871982 on Build GRCh38
rs369254539 Chr.19: 35872048-35872048 on Build GRCh38
rs962061258 Chr.19: 35872065-35872065 on Build GRCh38
rs200756165 Chr.19: 35871993-35871993 on Build GRCh38
rs766517343 Chr.19: 35872003-35872003 on Build GRCh38
rs760039233 Chr.19: 35872071-35872072 on Build GRCh38
rs373409117 Chr.19: 35872017-35872017 on Build GRCh38
rs745547684 Chr.19: 35871988-35871988 on Build GRCh38
rs111357652 Chr.19: 35872037-35872037 on Build GRCh38
rs772687634 Chr.19: 35872005-35872005 on Build GRCh38
rs892106146 Chr.19: 35871986-35871986 on Build GRCh38
rs944418640 Chr.19: 35871961-35871961 on Build GRCh38
rs566125112 Chr.19: 35872076-35872076 on Build GRCh38
rs951419368 Chr.19: 35872091-35872091 on Build GRCh38
rs376813918 Chr.19: 35871965-35871965 on Build GRCh38
rs113251170 Chr.19: 35872038-35872038 on Build GRCh38

View Full Product Details