Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111925751-111926014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf52
Entrez Gene ID
91894
Gene Name
chromosome 11 open reading frame 52
Gene Aliases
-
UniGene
Hs.97013
Gene Chromosome Location
Chr.11: 111918877-111926871 on Build GRCh38
Transcripts
NM_080659.2 NR_037651.1

Gene Symbol
HSPB2-C11orf52
Entrez Gene ID
100528019
Gene Name
HSPB2-C11orf52 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.97013
Gene Chromosome Location
Chr.11: 111912736-111926871 on Build GRCh38
Transcripts
NM_080659.2 NR_037651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4017837 c.163C>T p.R55W Chr.11: 111925990-111925990 on Build GRCh38
4017836 c.148G>T p.G50* Chr.11: 111925975-111925975 on Build GRCh38
1293305 c.139G>A p.E47K Chr.11: 111925966-111925966 on Build GRCh38
4989400 c.137A>G p.H46R Chr.11: 111925964-111925964 on Build GRCh38
2111846 c.139G>C p.E47Q Chr.11: 111925966-111925966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781792683 Chr.11: 111925964-111925964 on Build GRCh38
rs148069684 Chr.11: 111925941-111925941 on Build GRCh38
rs587603688 Chr.11: 111925852-111925852 on Build GRCh38
rs782333478 Chr.11: 111925900-111925900 on Build GRCh38
rs782002030 Chr.11: 111925952-111925952 on Build GRCh38
rs782714958 Chr.11: 111925942-111925943 on Build GRCh38
rs952111817 Chr.11: 111925991-111925991 on Build GRCh38
rs1007820968 Chr.11: 111925956-111925956 on Build GRCh38
rs117666512 Chr.11: 111925965-111925965 on Build GRCh38
rs917078768 Chr.11: 111925866-111925866 on Build GRCh38
rs782165448 Chr.11: 111925986-111925986 on Build GRCh38
rs782781686 Chr.11: 111925943-111925943 on Build GRCh38
rs587726693 Chr.11: 111925813-111925813 on Build GRCh38
rs782088394 Chr.11: 111925953-111925953 on Build GRCh38
rs949876679 Chr.11: 111925888-111925888 on Build GRCh38
rs782025291 Chr.11: 111925939-111925939 on Build GRCh38
rs782746133 Chr.11: 111925961-111925961 on Build GRCh38
rs370321292 Chr.11: 111925983-111925983 on Build GRCh38
rs781852399 Chr.11: 111925982-111925982 on Build GRCh38
rs937278053 Chr.11: 111925844-111925844 on Build GRCh38
rs1054106808 Chr.11: 111925938-111925938 on Build GRCh38
rs782333272 Chr.11: 111925931-111925931 on Build GRCh38
rs1055608641 Chr.11: 111925845-111925845 on Build GRCh38
rs587642593 Chr.11: 111925811-111925811 on Build GRCh38
rs115724940 Chr.11: 111925993-111925993 on Build GRCh38
rs782261294 Chr.11: 111925809-111925809 on Build GRCh38
rs147007936 Chr.11: 111925894-111925894 on Build GRCh38

View Full Product Details