Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 21002740-21002975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOB
Entrez Gene ID
338
Gene Name
apolipoprotein B
Gene Aliases
FLDB, LDLCQ4, apoB-100, apoB-48
UniGene
Hs.120759
Gene Chromosome Location
Chr.2: 21001429-21044073 on Build GRCh38
Transcripts
NM_000384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
339123 c.12526G>T p.V4176L Chr.2: 21002896-21002896 on Build GRCh38
3577120 c.12492C>T p.F4164F Chr.2: 21002930-21002930 on Build GRCh38
4524348 c.12674G>A p.G4225E Chr.2: 21002748-21002748 on Build GRCh38
3577119 c.12615G>A p.P4205P Chr.2: 21002807-21002807 on Build GRCh38
5396097 c.12667G>A p.E4223K Chr.2: 21002755-21002755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771559921 Chr.2: 21002888-21002888 on Build GRCh38
rs758266841 Chr.2: 21002874-21002874 on Build GRCh38
rs756670216 Chr.2: 21002869-21002869 on Build GRCh38
rs758599848 Chr.2: 21002828-21002828 on Build GRCh38
rs748744377 Chr.2: 21002782-21002782 on Build GRCh38
rs755234920 Chr.2: 21002842-21002842 on Build GRCh38
rs770139422 Chr.2: 21002783-21002783 on Build GRCh38
rs766149220 Chr.2: 21002810-21002810 on Build GRCh38
rs202247483 Chr.2: 21002807-21002807 on Build GRCh38
rs370180297 Chr.2: 21002886-21002886 on Build GRCh38
rs750537815 Chr.2: 21002825-21002825 on Build GRCh38
rs763896891 Chr.2: 21002817-21002817 on Build GRCh38
rs764649432 Chr.2: 21002912-21002912 on Build GRCh38
rs766535406 Chr.2: 21002835-21002835 on Build GRCh38
rs770408875 Chr.2: 21002795-21002795 on Build GRCh38
rs781402806 Chr.2: 21002778-21002778 on Build GRCh38
rs1042031 Chr.2: 21002881-21002881 on Build GRCh38
rs865916036 Chr.2: 21002779-21002779 on Build GRCh38
rs766623212 Chr.2: 21002939-21002939 on Build GRCh38
rs752965923 Chr.2: 21002861-21002861 on Build GRCh38
rs201208445 Chr.2: 21002787-21002787 on Build GRCh38
rs570782024 Chr.2: 21002841-21002841 on Build GRCh38
rs773475226 Chr.2: 21002843-21002844 on Build GRCh38
rs62122485 Chr.2: 21002947-21002947 on Build GRCh38
rs375275230 Chr.2: 21002893-21002893 on Build GRCh38
rs781627851 Chr.2: 21002855-21002855 on Build GRCh38
rs772812955 Chr.2: 21002786-21002786 on Build GRCh38
rs760668446 Chr.2: 21002813-21002813 on Build GRCh38
rs775384365 Chr.2: 21002890-21002890 on Build GRCh38
rs367740129 Chr.2: 21002879-21002879 on Build GRCh38
rs761838451 Chr.2: 21002938-21002938 on Build GRCh38
rs751074267 Chr.2: 21002871-21002871 on Build GRCh38
rs775091905 Chr.2: 21002812-21002812 on Build GRCh38
rs375471570 Chr.2: 21002808-21002808 on Build GRCh38
rs755429390 Chr.2: 21002766-21002766 on Build GRCh38
rs768472814 Chr.2: 21002911-21002911 on Build GRCh38
rs371191761 Chr.2: 21002884-21002884 on Build GRCh38

View Full Product Details