Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73502855-73503126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTF3
Entrez Gene ID
689
Gene Name
basic transcription factor 3
Gene Aliases
BETA-NAC, BTF3a, BTF3b, NACB
UniGene
Hs.591768
Gene Chromosome Location
Chr.5: 73498425-73505623 on Build GRCh38
Transcripts
NM_001037637.1 NM_001207.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069942 c.400A>G p.I134V Chr.5: 73503000-73503000 on Build GRCh38
3828392 c.473G>C p.S158T Chr.5: 73503073-73503073 on Build GRCh38
318960 c.508C>T p.P170S Chr.5: 73503108-73503108 on Build GRCh38
3828391 c.415G>C p.E139Q Chr.5: 73503015-73503015 on Build GRCh38
5447649 c.436A>G p.M146V Chr.5: 73503036-73503036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116176540 Chr.5: 73502884-73502884 on Build GRCh38
rs347233 Chr.5: 73502899-73502899 on Build GRCh38
rs143150590 Chr.5: 73503048-73503048 on Build GRCh38
rs966602011 Chr.5: 73502965-73502965 on Build GRCh38
rs754868948 Chr.5: 73502920-73502920 on Build GRCh38
rs778415639 Chr.5: 73502931-73502931 on Build GRCh38
rs186931485 Chr.5: 73503099-73503099 on Build GRCh38
rs746411488 Chr.5: 73502997-73502997 on Build GRCh38
rs951669972 Chr.5: 73502993-73502993 on Build GRCh38
rs768814807 Chr.5: 73503032-73503032 on Build GRCh38
rs764307835 Chr.5: 73502898-73502898 on Build GRCh38
rs760502541 Chr.5: 73503077-73503077 on Build GRCh38
rs111757484 Chr.5: 73502954-73502954 on Build GRCh38
rs770379410 Chr.5: 73503002-73503002 on Build GRCh38
rs763094367 Chr.5: 73502886-73502886 on Build GRCh38
rs151143923 Chr.5: 73502951-73502951 on Build GRCh38
rs41271745 Chr.5: 73503018-73503018 on Build GRCh38
rs752195686 Chr.5: 73502936-73502936 on Build GRCh38
rs1050429962 Chr.5: 73502937-73502937 on Build GRCh38
rs372460087 Chr.5: 73503067-73503067 on Build GRCh38
rs747392162 Chr.5: 73502944-73502944 on Build GRCh38
rs878981435 Chr.5: 73502962-73502962 on Build GRCh38
rs766351583 Chr.5: 73502908-73502908 on Build GRCh38
rs757993865 Chr.5: 73502940-73502940 on Build GRCh38
rs958788578 Chr.5: 73502911-73502911 on Build GRCh38
rs996682498 Chr.5: 73502922-73502922 on Build GRCh38
rs72768738 Chr.5: 73503068-73503068 on Build GRCh38
rs753743654 Chr.5: 73502912-73502912 on Build GRCh38
rs137911027 Chr.5: 73502987-73502987 on Build GRCh38
rs201235182 Chr.5: 73503049-73503049 on Build GRCh38
rs14010 Chr.5: 73503020-73503020 on Build GRCh38
rs753579016 Chr.5: 73503098-73503098 on Build GRCh38
rs201362870 Chr.5: 73503008-73503008 on Build GRCh38
rs766190108 Chr.5: 73503081-73503081 on Build GRCh38

View Full Product Details