Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12725890-12726159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOLD1
Entrez Gene ID
81575
Gene Name
apolipoprotein L domain containing 1
Gene Aliases
VERGE
UniGene
Hs.23388
Gene Chromosome Location
Chr.12: 12725917-12791466 on Build GRCh38
Transcripts
NM_001130415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7313623 c.3G>T p.M1I Chr.12: 12726003-12726003 on Build GRCh38
6239214 c.28C>T p.R10W Chr.12: 12726028-12726028 on Build GRCh38
4600806 c.29G>T p.R10L Chr.12: 12726029-12726029 on Build GRCh38
7417695 c.62C>G p.A21G Chr.12: 12726062-12726062 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs548492458 Chr.12: 12726042-12726042 on Build GRCh38
rs367605402 Chr.12: 12726087-12726087 on Build GRCh38
rs934813716 Chr.12: 12726023-12726023 on Build GRCh38
rs1038985043 Chr.12: 12726033-12726033 on Build GRCh38
rs1004451943 Chr.12: 12726117-12726117 on Build GRCh38
rs756679790 Chr.12: 12726108-12726109 on Build GRCh38
rs544203710 Chr.12: 12725950-12725950 on Build GRCh38
rs768784342 Chr.12: 12726046-12726046 on Build GRCh38
rs775679489 Chr.12: 12726039-12726039 on Build GRCh38
rs576829445 Chr.12: 12725936-12725936 on Build GRCh38
rs552054785 Chr.12: 12726131-12726131 on Build GRCh38
rs768053848 Chr.12: 12725936-12725937 on Build GRCh38
rs202116412 Chr.12: 12726097-12726097 on Build GRCh38
rs34323 Chr.12: 12726123-12726123 on Build GRCh38
rs987033485 Chr.12: 12725940-12725940 on Build GRCh38
rs904877617 Chr.12: 12726070-12726070 on Build GRCh38
rs7297629 Chr.12: 12726029-12726029 on Build GRCh38
rs781292749 Chr.12: 12725967-12725967 on Build GRCh38
rs909556771 Chr.12: 12725937-12725937 on Build GRCh38
rs770064762 Chr.12: 12726035-12726035 on Build GRCh38
rs767815596 Chr.12: 12726119-12726119 on Build GRCh38
rs560421199 Chr.12: 12726106-12726106 on Build GRCh38
rs757006225 Chr.12: 12725961-12725961 on Build GRCh38
rs562420626 Chr.12: 12726015-12726015 on Build GRCh38
rs946493506 Chr.12: 12726041-12726041 on Build GRCh38
rs565013547 Chr.12: 12725917-12725917 on Build GRCh38

View Full Product Details