Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 77294774-77295029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS18
Entrez Gene ID
170692
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 18
Gene Aliases
ADAMTS21, KNO2, MMCAT
UniGene
Hs.188746
Gene Chromosome Location
Chr.16: 77282128-77435114 on Build GRCh38
Transcripts
NM_001326358.1 NM_199355.3 XM_011522923.1 XM_011522924.1 XM_017022988.1 XM_017022989.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2155585 c.2964C>G p.S988R Chr.16: 77294965-77294965 on Build GRCh38
7291222 c.2928A>G p.P976P Chr.16: 77295001-77295001 on Build GRCh38
142245 c.3004C>T p.Q1002* Chr.16: 77294925-77294925 on Build GRCh38
4659529 c.2985G>T p.W995C Chr.16: 77294944-77294944 on Build GRCh38
352623 c.2953G>T p.A985S Chr.16: 77294976-77294976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763344799 Chr.16: 77295003-77295003 on Build GRCh38
rs750022707 Chr.16: 77294851-77294851 on Build GRCh38
rs745888861 Chr.16: 77294962-77294962 on Build GRCh38
rs771108884 Chr.16: 77294982-77294982 on Build GRCh38
rs755938429 Chr.16: 77294963-77294963 on Build GRCh38
rs747219080 Chr.16: 77294979-77294979 on Build GRCh38
rs772364406 Chr.16: 77294918-77294918 on Build GRCh38
rs755417562 Chr.16: 77294872-77294872 on Build GRCh38
rs183622474 Chr.16: 77294995-77294995 on Build GRCh38
rs371016487 Chr.16: 77294931-77294931 on Build GRCh38
rs201923014 Chr.16: 77294892-77294892 on Build GRCh38
rs561733123 Chr.16: 77294854-77294854 on Build GRCh38
rs191328145 Chr.16: 77294942-77294942 on Build GRCh38
rs751185373 Chr.16: 77294951-77294951 on Build GRCh38
rs759297566 Chr.16: 77294954-77294954 on Build GRCh38
rs895809075 Chr.16: 77294822-77294822 on Build GRCh38
rs373078127 Chr.16: 77294890-77294890 on Build GRCh38
rs979241407 Chr.16: 77294849-77294849 on Build GRCh38
rs760098320 Chr.16: 77294990-77294990 on Build GRCh38
rs776507473 Chr.16: 77294983-77294983 on Build GRCh38
rs757976342 Chr.16: 77294882-77294882 on Build GRCh38
rs759956055 Chr.16: 77294934-77294934 on Build GRCh38
rs201899506 Chr.16: 77294898-77294898 on Build GRCh38
rs987927627 Chr.16: 77294876-77294876 on Build GRCh38
rs779132393 Chr.16: 77294875-77294875 on Build GRCh38
rs779913181 Chr.16: 77294964-77294964 on Build GRCh38
rs918033771 Chr.16: 77294826-77294826 on Build GRCh38
rs763693273 Chr.16: 77294950-77294950 on Build GRCh38
rs144455612 Chr.16: 77294927-77294927 on Build GRCh38
rs745773712 Chr.16: 77294897-77294897 on Build GRCh38
rs748386211 Chr.16: 77294908-77294908 on Build GRCh38
rs756811678 Chr.16: 77294952-77294952 on Build GRCh38
rs992667185 Chr.16: 77294802-77294802 on Build GRCh38
rs969225751 Chr.16: 77294848-77294848 on Build GRCh38
rs775861888 Chr.16: 77295001-77295001 on Build GRCh38
rs958747943 Chr.16: 77294800-77294800 on Build GRCh38
rs201049858 Chr.16: 77294930-77294930 on Build GRCh38
rs775144991 Chr.16: 77294904-77294904 on Build GRCh38
rs141256982 Chr.16: 77294976-77294976 on Build GRCh38
rs1013471150 Chr.16: 77294827-77294827 on Build GRCh38
rs147793662 Chr.16: 77294975-77294975 on Build GRCh38
rs267604644 Chr.16: 77294932-77294932 on Build GRCh38
rs74584252 Chr.16: 77294969-77294969 on Build GRCh38
rs12935229 Chr.16: 77294998-77294998 on Build GRCh38
rs781233200 Chr.16: 77294893-77294893 on Build GRCh38
rs550367127 Chr.16: 77294940-77294940 on Build GRCh38
rs368865291 Chr.16: 77294959-77294959 on Build GRCh38
rs749044422 Chr.16: 77294965-77294965 on Build GRCh38
rs752461179 Chr.16: 77294881-77294881 on Build GRCh38
rs548207084 Chr.16: 77294828-77294828 on Build GRCh38
rs773449203 Chr.16: 77294924-77294924 on Build GRCh38

View Full Product Details