Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44071131-44071392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
UniGene
Hs.126706
Gene Chromosome Location
Chr.11: 44066118-44084237 on Build GRCh38
Transcripts
NM_001127219.1 NM_032592.3 XM_005253170.3 XM_005253172.2 XM_006718348.2 XM_011520406.2 XM_011520407.1 XM_011520410.2 XM_017018421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6591900 c.346C>T p.R116W Chr.11: 44071313-44071313 on Build GRCh38
1979172 c.309C>A p.T103T Chr.11: 44071276-44071276 on Build GRCh38
1675953 c.322C>T p.L108F Chr.11: 44071289-44071289 on Build GRCh38
3670764 c.347G>T p.R116L Chr.11: 44071314-44071314 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778975822 Chr.11: 44071277-44071277 on Build GRCh38
rs369107890 Chr.11: 44071248-44071248 on Build GRCh38
rs375840611 Chr.11: 44071214-44071214 on Build GRCh38
rs915770815 Chr.11: 44071206-44071206 on Build GRCh38
rs749247040 Chr.11: 44071259-44071259 on Build GRCh38
rs758836441 Chr.11: 44071215-44071215 on Build GRCh38
rs775734725 Chr.11: 44071287-44071287 on Build GRCh38
rs769773390 Chr.11: 44071234-44071234 on Build GRCh38
rs923602239 Chr.11: 44071293-44071293 on Build GRCh38
rs766917830 Chr.11: 44071341-44071341 on Build GRCh38
rs757681771 Chr.11: 44071212-44071212 on Build GRCh38
rs768589956 Chr.11: 44071291-44071291 on Build GRCh38
rs750950641 Chr.11: 44071227-44071228 on Build GRCh38
rs756747248 Chr.11: 44071238-44071239 on Build GRCh38
rs763468932 Chr.11: 44071338-44071338 on Build GRCh38
rs770955945 Chr.11: 44071267-44071267 on Build GRCh38
rs747749239 Chr.11: 44071229-44071229 on Build GRCh38
rs760859659 Chr.11: 44071289-44071289 on Build GRCh38
rs754420091 Chr.11: 44071220-44071220 on Build GRCh38
rs377581240 Chr.11: 44071225-44071225 on Build GRCh38
rs943163475 Chr.11: 44071274-44071274 on Build GRCh38
rs754580610 Chr.11: 44071353-44071353 on Build GRCh38
rs1039174524 Chr.11: 44071307-44071307 on Build GRCh38
rs780708148 Chr.11: 44071337-44071337 on Build GRCh38
rs750745642 Chr.11: 44071332-44071332 on Build GRCh38
rs946363667 Chr.11: 44071251-44071251 on Build GRCh38
rs780177608 Chr.11: 44071368-44071368 on Build GRCh38
rs915022231 Chr.11: 44071240-44071240 on Build GRCh38
rs757523897 Chr.11: 44071304-44071304 on Build GRCh38
rs748107746 Chr.11: 44071354-44071354 on Build GRCh38
rs761881371 Chr.11: 44071326-44071326 on Build GRCh38
rs981997551 Chr.11: 44071186-44071186 on Build GRCh38
rs1042029364 Chr.11: 44071256-44071256 on Build GRCh38
rs779221374 Chr.11: 44071313-44071313 on Build GRCh38
rs991743671 Chr.11: 44071182-44071182 on Build GRCh38
rs753988064 Chr.11: 44071207-44071207 on Build GRCh38
rs927768598 Chr.11: 44071197-44071197 on Build GRCh38
rs763742730 Chr.11: 44071212-44071212 on Build GRCh38
rs2074039 Chr.11: 44071330-44071330 on Build GRCh38
rs754253971 Chr.11: 44071166-44071166 on Build GRCh38
rs751976387 Chr.11: 44071350-44071350 on Build GRCh38
rs373156903 Chr.11: 44071250-44071250 on Build GRCh38
rs139319943 Chr.11: 44071217-44071217 on Build GRCh38
rs150764409 Chr.11: 44071176-44071176 on Build GRCh38

View Full Product Details