Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20419255-20419514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM5
Entrez Gene ID
54988
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
-
UniGene
Hs.659606
Gene Chromosome Location
Chr.16: 20409508-20441333 on Build GRCh38
Transcripts
NM_001324371.1 NM_001324372.1 NM_001324373.1 NM_017888.2 XM_006721056.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297027 c.465G>A p.K155K Chr.16: 20419277-20419277 on Build GRCh38
4974051 c.514C>G p.L172V Chr.16: 20419326-20419326 on Build GRCh38
5706033 c.551C>T p.P184L Chr.16: 20419363-20419363 on Build GRCh38
5690270 c.446T>A p.L149Q Chr.16: 20419258-20419258 on Build GRCh38
6502036 c.501C>G p.I167M Chr.16: 20419313-20419313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774633444 Chr.16: 20419469-20419469 on Build GRCh38
rs771212949 Chr.16: 20419395-20419395 on Build GRCh38
rs745696906 Chr.16: 20419373-20419373 on Build GRCh38
rs774489244 Chr.16: 20419400-20419400 on Build GRCh38
rs531998936 Chr.16: 20419294-20419294 on Build GRCh38
rs144928537 Chr.16: 20419361-20419361 on Build GRCh38
rs778532803 Chr.16: 20419371-20419371 on Build GRCh38
rs770959278 Chr.16: 20419463-20419463 on Build GRCh38
rs139646135 Chr.16: 20419275-20419275 on Build GRCh38
rs1052165971 Chr.16: 20419391-20419391 on Build GRCh38
rs759917214 Chr.16: 20419351-20419351 on Build GRCh38
rs149196840 Chr.16: 20419435-20419435 on Build GRCh38
rs756938890 Chr.16: 20419370-20419370 on Build GRCh38
rs749010470 Chr.16: 20419288-20419288 on Build GRCh38
rs1000728324 Chr.16: 20419378-20419378 on Build GRCh38
rs374023853 Chr.16: 20419310-20419310 on Build GRCh38
rs528643775 Chr.16: 20419295-20419295 on Build GRCh38
rs767839776 Chr.16: 20419402-20419402 on Build GRCh38
rs144504078 Chr.16: 20419291-20419291 on Build GRCh38
rs567568290 Chr.16: 20419482-20419482 on Build GRCh38
rs572015052 Chr.16: 20419282-20419282 on Build GRCh38
rs370016772 Chr.16: 20419480-20419480 on Build GRCh38
rs139829456 Chr.16: 20419341-20419341 on Build GRCh38
rs563078080 Chr.16: 20419381-20419381 on Build GRCh38
rs77322809 Chr.16: 20419447-20419447 on Build GRCh38
rs761961501 Chr.16: 20419332-20419332 on Build GRCh38
rs761131750 Chr.16: 20419488-20419488 on Build GRCh38
rs751953070 Chr.16: 20419347-20419347 on Build GRCh38
rs754967486 Chr.16: 20419457-20419457 on Build GRCh38
rs200884260 Chr.16: 20419441-20419441 on Build GRCh38
rs775966359 Chr.16: 20419423-20419423 on Build GRCh38
rs201728148 Chr.16: 20419354-20419354 on Build GRCh38
rs7192210 Chr.16: 20419356-20419356 on Build GRCh38
rs771811390 Chr.16: 20419376-20419376 on Build GRCh38
rs4072768 Chr.16: 20419446-20419446 on Build GRCh38
rs780051706 Chr.16: 20419449-20419449 on Build GRCh38
rs186088988 Chr.16: 20419355-20419355 on Build GRCh38
rs112760108 Chr.16: 20419352-20419352 on Build GRCh38
rs375729525 Chr.16: 20419462-20419462 on Build GRCh38
rs372288147 Chr.16: 20419475-20419475 on Build GRCh38
rs755518647 Chr.16: 20419277-20419277 on Build GRCh38
rs559741756 Chr.16: 20419289-20419289 on Build GRCh38
rs533410184 Chr.16: 20419434-20419434 on Build GRCh38
rs746881941 Chr.16: 20419453-20419453 on Build GRCh38
rs373907643 Chr.16: 20419401-20419401 on Build GRCh38
rs202200151 Chr.16: 20419452-20419453 on Build GRCh38
rs549370944 Chr.16: 20419481-20419481 on Build GRCh38
rs777956169 Chr.16: 20419460-20419460 on Build GRCh38
rs201001714 Chr.16: 20419281-20419281 on Build GRCh38
rs372626063 Chr.16: 20419304-20419304 on Build GRCh38
rs761185237 Chr.16: 20419424-20419424 on Build GRCh38
rs778784463 Chr.16: 20419292-20419292 on Build GRCh38
rs797013215 Chr.16: 20419352-20419353 on Build GRCh38
rs776524157 Chr.16: 20419313-20419313 on Build GRCh38
rs773436882 Chr.16: 20419382-20419382 on Build GRCh38
rs1057075317 Chr.16: 20419343-20419343 on Build GRCh38
rs770031289 Chr.16: 20419337-20419337 on Build GRCh38
rs747008520 Chr.16: 20419377-20419377 on Build GRCh38
rs749400338 Chr.16: 20419383-20419383 on Build GRCh38

View Full Product Details