Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4718398-4718641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK3
Entrez Gene ID
50808
Gene Name
adenylate kinase 3
Gene Aliases
AK3L1, AK6, AKL3L, AKL3L1, FIX
UniGene
Hs.732022
Gene Chromosome Location
Chr.9: 4709556-4742043 on Build GRCh38
Transcripts
NM_001199852.1 NM_001199853.1 NM_001199855.1 NM_001199856.1 NM_016282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6843592 c.475A>T p.I159F Chr.9: 4718507-4718507 on Build GRCh38
3907505 c.504G>A p.T168T Chr.9: 4718478-4718478 on Build GRCh38
69596 c.535C>G p.Q179E Chr.9: 4718447-4718447 on Build GRCh38
2777150 c.503C>T p.T168M Chr.9: 4718479-4718479 on Build GRCh38
293356 c.456C>G p.D152E Chr.9: 4718526-4718526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771116247 Chr.9: 4718586-4718586 on Build GRCh38
rs146879241 Chr.9: 4718520-4718520 on Build GRCh38
rs151087607 Chr.9: 4718440-4718440 on Build GRCh38
rs575116386 Chr.9: 4718606-4718606 on Build GRCh38
rs372927818 Chr.9: 4718553-4718553 on Build GRCh38
rs183973361 Chr.9: 4718563-4718563 on Build GRCh38
rs371256594 Chr.9: 4718587-4718587 on Build GRCh38
rs374515313 Chr.9: 4718554-4718554 on Build GRCh38
rs748671719 Chr.9: 4718471-4718471 on Build GRCh38
rs773431848 Chr.9: 4718511-4718511 on Build GRCh38
rs773391830 Chr.9: 4718448-4718448 on Build GRCh38
rs745431034 Chr.9: 4718556-4718556 on Build GRCh38
rs953055512 Chr.9: 4718501-4718501 on Build GRCh38
rs758722360 Chr.9: 4718543-4718543 on Build GRCh38
rs768600522 Chr.9: 4718507-4718507 on Build GRCh38
rs764466177 Chr.9: 4718546-4718546 on Build GRCh38
rs369685256 Chr.9: 4718443-4718443 on Build GRCh38
rs147962892 Chr.9: 4718529-4718529 on Build GRCh38
rs570611598 Chr.9: 4718485-4718485 on Build GRCh38
rs572769835 Chr.9: 4718574-4718574 on Build GRCh38
rs753029873 Chr.9: 4718539-4718539 on Build GRCh38
rs766413945 Chr.9: 4718519-4718519 on Build GRCh38
rs760817849 Chr.9: 4718514-4718514 on Build GRCh38
rs749257216 Chr.9: 4718570-4718570 on Build GRCh38
rs188927307 Chr.9: 4718588-4718588 on Build GRCh38
rs201566150 Chr.9: 4718481-4718481 on Build GRCh38
rs917753302 Chr.9: 4718595-4718595 on Build GRCh38
rs772482535 Chr.9: 4718562-4718563 on Build GRCh38
rs141049546 Chr.9: 4718478-4718478 on Build GRCh38
rs769922742 Chr.9: 4718503-4718503 on Build GRCh38
rs946955968 Chr.9: 4718605-4718605 on Build GRCh38
rs375214094 Chr.9: 4718500-4718500 on Build GRCh38
rs768510696 Chr.9: 4718571-4718571 on Build GRCh38
rs751167622 Chr.9: 4718549-4718549 on Build GRCh38
rs144768148 Chr.9: 4718504-4718504 on Build GRCh38
rs866591478 Chr.9: 4718432-4718432 on Build GRCh38
rs555051720 Chr.9: 4718604-4718604 on Build GRCh38
rs949275443 Chr.9: 4718598-4718598 on Build GRCh38
rs1044977623 Chr.9: 4718620-4718620 on Build GRCh38
rs975689071 Chr.9: 4718486-4718486 on Build GRCh38
rs767912598 Chr.9: 4718446-4718446 on Build GRCh38
rs749131195 Chr.9: 4718506-4718506 on Build GRCh38
rs915191986 Chr.9: 4718599-4718599 on Build GRCh38
rs756744534 Chr.9: 4718498-4718498 on Build GRCh38
rs760763787 Chr.9: 4718450-4718450 on Build GRCh38
rs770499972 Chr.9: 4718593-4718593 on Build GRCh38
rs770863203 Chr.9: 4718466-4718466 on Build GRCh38
rs780095261 Chr.9: 4718568-4718568 on Build GRCh38
rs887211999 Chr.9: 4718433-4718433 on Build GRCh38
rs747248787 Chr.9: 4718585-4718585 on Build GRCh38
rs758023810 Chr.9: 4718479-4718479 on Build GRCh38
rs766390444 Chr.9: 4718460-4718460 on Build GRCh38
rs529055631 Chr.9: 4718496-4718496 on Build GRCh38
rs765581630 Chr.9: 4718477-4718477 on Build GRCh38

View Full Product Details