Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22976275-22976546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AJUBA
Entrez Gene ID
84962
Gene Name
ajuba LIM protein
Gene Aliases
JUB
UniGene
Hs.655832
Gene Chromosome Location
Chr.14: 22971174-22982642 on Build GRCh38
Transcripts
NM_032876.5 NM_198086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5632871 c.1216T>A p.C406S Chr.14: 22976479-22976479 on Build GRCh38
6075572 c.1186T>A p.F396I Chr.14: 22976509-22976509 on Build GRCh38
3793579 c.1201G>C p.E401Q Chr.14: 22976494-22976494 on Build GRCh38
6867500 c.1239+1G>A p.? Chr.14: 22976455-22976455 on Build GRCh38
1493210 c.1177-2A>T p.? Chr.14: 22976520-22976520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020714604 Chr.14: 22976422-22976422 on Build GRCh38
rs771154698 Chr.14: 22976342-22976350 on Build GRCh38
rs758514551 Chr.14: 22976327-22976327 on Build GRCh38
rs887079993 Chr.14: 22976367-22976367 on Build GRCh38
rs184637137 Chr.14: 22976416-22976416 on Build GRCh38
rs747184863 Chr.14: 22976328-22976328 on Build GRCh38
rs759310932 Chr.14: 22976406-22976406 on Build GRCh38
rs367814359 Chr.14: 22976417-22976417 on Build GRCh38
rs780714755 Chr.14: 22976459-22976459 on Build GRCh38
rs770057260 Chr.14: 22976523-22976523 on Build GRCh38
rs757036893 Chr.14: 22976343-22976343 on Build GRCh38
rs189945692 Chr.14: 22976359-22976359 on Build GRCh38
rs546140531 Chr.14: 22976510-22976510 on Build GRCh38
rs10134257 Chr.14: 22976327-22976327 on Build GRCh38
rs115048774 Chr.14: 22976332-22976332 on Build GRCh38
rs758781837 Chr.14: 22976449-22976449 on Build GRCh38
rs1004236301 Chr.14: 22976373-22976373 on Build GRCh38
rs201454214 Chr.14: 22976481-22976481 on Build GRCh38
rs1029775475 Chr.14: 22976400-22976400 on Build GRCh38
rs1009877968 Chr.14: 22976378-22976378 on Build GRCh38
rs1020248082 Chr.14: 22976388-22976388 on Build GRCh38
rs750877864 Chr.14: 22976448-22976448 on Build GRCh38
rs372729466 Chr.14: 22976424-22976424 on Build GRCh38
rs755108806 Chr.14: 22976499-22976499 on Build GRCh38
rs781179626 Chr.14: 22976521-22976521 on Build GRCh38
rs764974413 Chr.14: 22976433-22976433 on Build GRCh38
rs767453818 Chr.14: 22976407-22976407 on Build GRCh38
rs760263623 Chr.14: 22976413-22976413 on Build GRCh38
rs998767693 Chr.14: 22976395-22976395 on Build GRCh38
rs751476496 Chr.14: 22976333-22976333 on Build GRCh38
rs370274837 Chr.14: 22976391-22976391 on Build GRCh38
rs1054148833 Chr.14: 22976340-22976340 on Build GRCh38
rs192294093 Chr.14: 22976390-22976390 on Build GRCh38
rs866960243 Chr.14: 22976511-22976511 on Build GRCh38
rs748229445 Chr.14: 22976522-22976522 on Build GRCh38
rs376393546 Chr.14: 22976410-22976410 on Build GRCh38
rs747434959 Chr.14: 22976465-22976465 on Build GRCh38
rs954166158 Chr.14: 22976404-22976404 on Build GRCh38

View Full Product Details