Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138223238-138223477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
UniGene
Hs.744868
Gene Chromosome Location
Chr.3: 138187248-138298389 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_014154.4 NM_015396.5 NM_213654.3 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 XM_017006100.1 XM_017006101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604990 c.282T>C p.T94T Chr.3: 138223476-138223476 on Build GRCh38
5987717 c.182C>G p.S61* Chr.3: 138223418-138223418 on Build GRCh38
1183644 c.185G>A p.S62N Chr.3: 138223421-138223421 on Build GRCh38
5987716 c.182C>G p.S61* Chr.3: 138223418-138223418 on Build GRCh38
5560131 c.226C>A p.L76I Chr.3: 138223462-138223462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002114829 Chr.3: 138223271-138223271 on Build GRCh38
rs771094197 Chr.3: 138223383-138223383 on Build GRCh38
rs557004033 Chr.3: 138223373-138223373 on Build GRCh38
rs200183090 Chr.3: 138223364-138223364 on Build GRCh38
rs149623348 Chr.3: 138223282-138223282 on Build GRCh38
rs769564958 Chr.3: 138223365-138223365 on Build GRCh38
rs774244966 Chr.3: 138223407-138223407 on Build GRCh38
rs764544651 Chr.3: 138223442-138223442 on Build GRCh38
rs754407141 Chr.3: 138223448-138223448 on Build GRCh38
rs115045683 Chr.3: 138223284-138223284 on Build GRCh38
rs968136063 Chr.3: 138223291-138223291 on Build GRCh38
rs987659705 Chr.3: 138223329-138223329 on Build GRCh38
rs979843873 Chr.3: 138223310-138223310 on Build GRCh38
rs576805045 Chr.3: 138223402-138223402 on Build GRCh38
rs761176368 Chr.3: 138223437-138223437 on Build GRCh38
rs924090457 Chr.3: 138223334-138223334 on Build GRCh38
rs910517204 Chr.3: 138223315-138223315 on Build GRCh38
rs954874448 Chr.3: 138223307-138223307 on Build GRCh38
rs1021468156 Chr.3: 138223287-138223287 on Build GRCh38
rs767781229 Chr.3: 138223422-138223422 on Build GRCh38
rs1034888414 Chr.3: 138223273-138223273 on Build GRCh38
rs375651235 Chr.3: 138223358-138223358 on Build GRCh38
rs568642456 Chr.3: 138223308-138223308 on Build GRCh38
rs539418203 Chr.3: 138223415-138223415 on Build GRCh38
rs146088572 Chr.3: 138223423-138223423 on Build GRCh38
rs1029219570 Chr.3: 138223304-138223304 on Build GRCh38

View Full Product Details