Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24318557-24318810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY4
Entrez Gene ID
196883
Gene Name
adenylate cyclase 4
Gene Aliases
AC4
UniGene
Hs.443428
Gene Chromosome Location
Chr.14: 24318349-24335071 on Build GRCh38
Transcripts
NM_001198568.1 NM_001198592.1 NM_139247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6898074 c.3029C>T p.T1010I Chr.14: 24318706-24318706 on Build GRCh38
6594940 c.3035A>G p.N1012S Chr.14: 24318700-24318700 on Build GRCh38
5382149 c.3082G>A p.V1028M Chr.14: 24318568-24318568 on Build GRCh38
3885950 c.3066C>T p.V1022V Chr.14: 24318669-24318669 on Build GRCh38
6308884 c.3082G>C p.V1028L Chr.14: 24318568-24318568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77376311 Chr.14: 24318582-24318582 on Build GRCh38
rs748416816 Chr.14: 24318647-24318647 on Build GRCh38
rs750646970 Chr.14: 24318585-24318585 on Build GRCh38
rs151266711 Chr.14: 24318688-24318688 on Build GRCh38
rs564894633 Chr.14: 24318768-24318768 on Build GRCh38
rs769107327 Chr.14: 24318641-24318641 on Build GRCh38
rs372811153 Chr.14: 24318607-24318607 on Build GRCh38
rs757989927 Chr.14: 24318745-24318745 on Build GRCh38
rs777261351 Chr.14: 24318644-24318644 on Build GRCh38
rs143391430 Chr.14: 24318675-24318675 on Build GRCh38
rs373039093 Chr.14: 24318586-24318586 on Build GRCh38
rs753753839 Chr.14: 24318720-24318720 on Build GRCh38
rs773676105 Chr.14: 24318683-24318683 on Build GRCh38
rs757144908 Chr.14: 24318729-24318729 on Build GRCh38
rs780032356 Chr.14: 24318599-24318599 on Build GRCh38
rs756021946 Chr.14: 24318605-24318605 on Build GRCh38
rs34455539 Chr.14: 24318596-24318597 on Build GRCh38
rs199619931 Chr.14: 24318627-24318627 on Build GRCh38
rs375782299 Chr.14: 24318626-24318626 on Build GRCh38
rs781556710 Chr.14: 24318632-24318632 on Build GRCh38
rs763623868 Chr.14: 24318687-24318687 on Build GRCh38
rs766567451 Chr.14: 24318698-24318698 on Build GRCh38
rs376677760 Chr.14: 24318580-24318580 on Build GRCh38
rs1010672535 Chr.14: 24318618-24318618 on Build GRCh38
rs756898834 Chr.14: 24318743-24318743 on Build GRCh38
rs961093269 Chr.14: 24318621-24318621 on Build GRCh38
rs370081321 Chr.14: 24318762-24318762 on Build GRCh38
rs755084061 Chr.14: 24318705-24318705 on Build GRCh38
rs751597622 Chr.14: 24318699-24318699 on Build GRCh38
rs910946971 Chr.14: 24318785-24318785 on Build GRCh38
rs1022671396 Chr.14: 24318587-24318587 on Build GRCh38
rs199633085 Chr.14: 24318628-24318628 on Build GRCh38
rs764077343 Chr.14: 24318706-24318706 on Build GRCh38
rs779749774 Chr.14: 24318761-24318761 on Build GRCh38
rs748700723 Chr.14: 24318776-24318776 on Build GRCh38
rs749148786 Chr.14: 24318624-24318624 on Build GRCh38
rs765662037 Chr.14: 24318682-24318682 on Build GRCh38
rs770030440 Chr.14: 24318657-24318657 on Build GRCh38
rs770700861 Chr.14: 24318625-24318625 on Build GRCh38
rs760557441 Chr.14: 24318600-24318601 on Build GRCh38
rs751611683 Chr.14: 24318602-24318602 on Build GRCh38
rs150391939 Chr.14: 24318730-24318730 on Build GRCh38
rs1001629291 Chr.14: 24318584-24318584 on Build GRCh38

View Full Product Details