Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110042252-110042513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf76
Entrez Gene ID
400073
Gene Name
chromosome 12 open reading frame 76
Gene Aliases
-
UniGene
Hs.44817
Gene Chromosome Location
Chr.12: 110041177-110067695 on Build GRCh38
Transcripts
NM_207435.2 XM_017019297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285987 c.382G>T p.V128L Chr.12: 110042377-110042377 on Build GRCh38
118431 c.378G>A p.A126A Chr.12: 110042381-110042381 on Build GRCh38
691301 c.309G>A p.G103G Chr.12: 110042450-110042450 on Build GRCh38
934911 c.377C>T p.A126V Chr.12: 110042382-110042382 on Build GRCh38
5456257 c.406T>C p.*136R Chr.12: 110042353-110042353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888170138 Chr.12: 110042325-110042325 on Build GRCh38
rs763306706 Chr.12: 110042430-110042430 on Build GRCh38
rs202016063 Chr.12: 110042388-110042388 on Build GRCh38
rs776311969 Chr.12: 110042464-110042464 on Build GRCh38
rs747693711 Chr.12: 110042335-110042335 on Build GRCh38
rs1022966610 Chr.12: 110042455-110042455 on Build GRCh38
rs904677727 Chr.12: 110042337-110042337 on Build GRCh38
rs753073008 Chr.12: 110042486-110042487 on Build GRCh38
rs368304966 Chr.12: 110042375-110042375 on Build GRCh38
rs1045248266 Chr.12: 110042352-110042352 on Build GRCh38
rs776964702 Chr.12: 110042385-110042385 on Build GRCh38
rs757871041 Chr.12: 110042305-110042305 on Build GRCh38
rs754568927 Chr.12: 110042323-110042323 on Build GRCh38
rs904854704 Chr.12: 110042369-110042369 on Build GRCh38
rs140080646 Chr.12: 110042461-110042461 on Build GRCh38
rs11550081 Chr.12: 110042313-110042313 on Build GRCh38
rs374404314 Chr.12: 110042331-110042331 on Build GRCh38
rs372750833 Chr.12: 110042301-110042301 on Build GRCh38
rs746636629 Chr.12: 110042378-110042378 on Build GRCh38
rs746682172 Chr.12: 110042317-110042317 on Build GRCh38
rs755301642 Chr.12: 110042438-110042438 on Build GRCh38
rs770659586 Chr.12: 110042449-110042449 on Build GRCh38
rs934838302 Chr.12: 110042380-110042380 on Build GRCh38
rs780869453 Chr.12: 110042440-110042440 on Build GRCh38
rs764372891 Chr.12: 110042382-110042382 on Build GRCh38
rs146562405 Chr.12: 110042433-110042433 on Build GRCh38
rs1035906290 Chr.12: 110042379-110042379 on Build GRCh38
rs780281581 Chr.12: 110042315-110042315 on Build GRCh38
rs751824718 Chr.12: 110042435-110042435 on Build GRCh38
rs555325986 Chr.12: 110042338-110042338 on Build GRCh38
rs370206147 Chr.12: 110042350-110042350 on Build GRCh38
rs770763405 Chr.12: 110042370-110042370 on Build GRCh38
rs765396544 Chr.12: 110042405-110042405 on Build GRCh38
rs765886542 Chr.12: 110042306-110042306 on Build GRCh38
rs749174511 Chr.12: 110042354-110042354 on Build GRCh38
rs751009249 Chr.12: 110042425-110042425 on Build GRCh38
rs371803112 Chr.12: 110042381-110042381 on Build GRCh38

View Full Product Details