Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36441029-36441298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNA1
Entrez Gene ID
8900
Gene Name
cyclin A1
Gene Aliases
CT146
UniGene
Hs.417050
Gene Chromosome Location
Chr.13: 36430488-36442882 on Build GRCh38
Transcripts
NM_001111045.1 NM_001111046.1 NM_001111047.1 NM_003914.3 XM_011535294.2 XM_011535295.2 XM_011535296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
118326 c.1173T>A p.A391A Chr.13: 36441192-36441192 on Build GRCh38
3360080 c.1202A>G p.K401R Chr.13: 36441221-36441221 on Build GRCh38
2072574 c.1132C>T p.P378S Chr.13: 36441151-36441151 on Build GRCh38
2072575 c.1203G>C p.K401N Chr.13: 36441222-36441222 on Build GRCh38
6074756 c.1129G>C p.D377H Chr.13: 36441148-36441148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145699331 Chr.13: 36441163-36441163 on Build GRCh38
rs751103800 Chr.13: 36441071-36441071 on Build GRCh38
rs763523941 Chr.13: 36441070-36441070 on Build GRCh38
rs114854781 Chr.13: 36441267-36441267 on Build GRCh38
rs780127979 Chr.13: 36441113-36441113 on Build GRCh38
rs749179710 Chr.13: 36441121-36441121 on Build GRCh38
rs756349933 Chr.13: 36441271-36441271 on Build GRCh38
rs767692772 Chr.13: 36441134-36441134 on Build GRCh38
rs974177249 Chr.13: 36441222-36441222 on Build GRCh38
rs771151854 Chr.13: 36441234-36441234 on Build GRCh38
rs1015130861 Chr.13: 36441120-36441120 on Build GRCh38
rs981753781 Chr.13: 36441266-36441266 on Build GRCh38
rs756889569 Chr.13: 36441079-36441079 on Build GRCh38
rs905746973 Chr.13: 36441094-36441094 on Build GRCh38
rs776781987 Chr.13: 36441265-36441265 on Build GRCh38
rs180874885 Chr.13: 36441174-36441174 on Build GRCh38
rs376205726 Chr.13: 36441085-36441085 on Build GRCh38
rs371483597 Chr.13: 36441069-36441069 on Build GRCh38
rs899717975 Chr.13: 36441066-36441066 on Build GRCh38
rs368269809 Chr.13: 36441093-36441093 on Build GRCh38
rs948281629 Chr.13: 36441220-36441220 on Build GRCh38
rs369777099 Chr.13: 36441176-36441176 on Build GRCh38
rs17188012 Chr.13: 36441105-36441105 on Build GRCh38
rs146743791 Chr.13: 36441177-36441177 on Build GRCh38
rs557574238 Chr.13: 36441058-36441058 on Build GRCh38
rs769275091 Chr.13: 36441154-36441156 on Build GRCh38
rs1035323198 Chr.13: 36441108-36441108 on Build GRCh38
rs963194750 Chr.13: 36441211-36441211 on Build GRCh38

View Full Product Details