Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83858705-83858978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL3
Entrez Gene ID
57188
Gene Name
ADAMTS like 3
Gene Aliases
ADAMTSL-3
UniGene
Hs.459162
Gene Chromosome Location
Chr.15: 83654086-84039842 on Build GRCh38
Transcripts
NM_001301110.1 NM_207517.2 XM_011521822.2 XM_011521823.2 XM_011521824.2 XM_011521825.2 XM_017022434.1 XM_017022435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6143338 c.773G>A p.R258K Chr.15: 83858811-83858811 on Build GRCh38
7101003 c.764G>A p.R255Q Chr.15: 83858802-83858802 on Build GRCh38
1375030 c.733G>T p.E245* Chr.15: 83858771-83858771 on Build GRCh38
5383958 c.758G>A p.G253E Chr.15: 83858796-83858796 on Build GRCh38
965854 c.746C>A p.A249D Chr.15: 83858784-83858784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375928569 Chr.15: 83858789-83858789 on Build GRCh38
rs754592805 Chr.15: 83858767-83858767 on Build GRCh38
rs755672773 Chr.15: 83858758-83858758 on Build GRCh38
rs748607535 Chr.15: 83858782-83858782 on Build GRCh38
rs978952935 Chr.15: 83858938-83858938 on Build GRCh38
rs759780317 Chr.15: 83858734-83858734 on Build GRCh38
rs200415584 Chr.15: 83858859-83858859 on Build GRCh38
rs779492532 Chr.15: 83858762-83858762 on Build GRCh38
rs753287669 Chr.15: 83858888-83858888 on Build GRCh38
rs779162812 Chr.15: 83858773-83858773 on Build GRCh38
rs761733346 Chr.15: 83858833-83858833 on Build GRCh38
rs972080699 Chr.15: 83858901-83858901 on Build GRCh38
rs74606988 Chr.15: 83858778-83858778 on Build GRCh38
rs773157770 Chr.15: 83858871-83858871 on Build GRCh38
rs149710156 Chr.15: 83858802-83858802 on Build GRCh38
rs142508012 Chr.15: 83858909-83858910 on Build GRCh38
rs377263020 Chr.15: 83858740-83858740 on Build GRCh38
rs770000702 Chr.15: 83858824-83858824 on Build GRCh38
rs530648675 Chr.15: 83858903-83858903 on Build GRCh38
rs149499512 Chr.15: 83858757-83858757 on Build GRCh38
rs771154300 Chr.15: 83858801-83858801 on Build GRCh38
rs571695559 Chr.15: 83858924-83858924 on Build GRCh38
rs367777513 Chr.15: 83858878-83858878 on Build GRCh38
rs959764432 Chr.15: 83858893-83858894 on Build GRCh38
rs370791528 Chr.15: 83858748-83858748 on Build GRCh38
rs775560578 Chr.15: 83858828-83858828 on Build GRCh38
rs778387761 Chr.15: 83858796-83858796 on Build GRCh38
rs747494720 Chr.15: 83858799-83858799 on Build GRCh38
rs370536321 Chr.15: 83858803-83858803 on Build GRCh38
rs184576391 Chr.15: 83858940-83858940 on Build GRCh38
rs767546793 Chr.15: 83858749-83858749 on Build GRCh38
rs371393930 Chr.15: 83858881-83858881 on Build GRCh38
rs746087824 Chr.15: 83858819-83858819 on Build GRCh38
rs767183344 Chr.15: 83858856-83858856 on Build GRCh38
rs753521217 Chr.15: 83858763-83858763 on Build GRCh38
rs751302187 Chr.15: 83858744-83858744 on Build GRCh38

View Full Product Details