Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43848733-43848992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300LG
Entrez Gene ID
146894
Gene Name
CD300 molecule like family member g
Gene Aliases
CLM-9, CLM9, NEPMUCIN, TREM-4, TREM4
UniGene
Hs.147313
Gene Chromosome Location
Chr.17: 43846777-43863639 on Build GRCh38
Transcripts
NM_001168322.1 NM_001168323.1 NM_001168324.1 NM_145273.3 XM_005257075.2 XM_005257076.2 XM_006721721.2 XM_006721722.2 XM_011524384.2 XM_017024235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4579738 c.372C>T p.V124V Chr.17: 43848886-43848886 on Build GRCh38
1710345 c.328G>A p.E110K Chr.17: 43848842-43848842 on Build GRCh38
4617770 c.303C>T p.D101D Chr.17: 43848817-43848817 on Build GRCh38
5906046 c.317G>A p.W106* Chr.17: 43848831-43848831 on Build GRCh38
5451444 c.314A>G p.Y105C Chr.17: 43848828-43848828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570779213 Chr.17: 43848832-43848832 on Build GRCh38
rs1049713176 Chr.17: 43848781-43848781 on Build GRCh38
rs750433045 Chr.17: 43848898-43848898 on Build GRCh38
rs147596228 Chr.17: 43848886-43848886 on Build GRCh38
rs111842445 Chr.17: 43848895-43848895 on Build GRCh38
rs200503531 Chr.17: 43848875-43848875 on Build GRCh38
rs925663648 Chr.17: 43848952-43848952 on Build GRCh38
rs145473485 Chr.17: 43848759-43848759 on Build GRCh38
rs374785527 Chr.17: 43848817-43848817 on Build GRCh38
rs756194750 Chr.17: 43848901-43848901 on Build GRCh38
rs113310194 Chr.17: 43848897-43848897 on Build GRCh38
rs978399985 Chr.17: 43848954-43848954 on Build GRCh38
rs754763095 Chr.17: 43848925-43848925 on Build GRCh38
rs778614514 Chr.17: 43848941-43848941 on Build GRCh38
rs765674061 Chr.17: 43848818-43848818 on Build GRCh38
rs372637400 Chr.17: 43848860-43848860 on Build GRCh38
rs992784546 Chr.17: 43848885-43848885 on Build GRCh38
rs904244227 Chr.17: 43848767-43848767 on Build GRCh38
rs778809970 Chr.17: 43848857-43848857 on Build GRCh38
rs148841815 Chr.17: 43848768-43848768 on Build GRCh38
rs774280106 Chr.17: 43848780-43848780 on Build GRCh38
rs367773909 Chr.17: 43848859-43848859 on Build GRCh38
rs201215891 Chr.17: 43848900-43848900 on Build GRCh38
rs770665682 Chr.17: 43848881-43848881 on Build GRCh38
rs892715932 Chr.17: 43848801-43848805 on Build GRCh38
rs548690727 Chr.17: 43848930-43848930 on Build GRCh38
rs778447487 Chr.17: 43848757-43848757 on Build GRCh38
rs754785243 Chr.17: 43848856-43848856 on Build GRCh38
rs997810107 Chr.17: 43848808-43848808 on Build GRCh38
rs772648347 Chr.17: 43848873-43848873 on Build GRCh38
rs752017257 Chr.17: 43848838-43848838 on Build GRCh38
rs767557360 Chr.17: 43848842-43848842 on Build GRCh38
rs145115383 Chr.17: 43848841-43848841 on Build GRCh38
rs556684443 Chr.17: 43848890-43848890 on Build GRCh38
rs766533868 Chr.17: 43848903-43848903 on Build GRCh38
rs1016739129 Chr.17: 43848846-43848846 on Build GRCh38
rs749105826 Chr.17: 43848774-43848774 on Build GRCh38
rs771541321 Chr.17: 43848786-43848786 on Build GRCh38
rs756282737 Chr.17: 43848849-43848849 on Build GRCh38
rs772916001 Chr.17: 43848811-43848811 on Build GRCh38
rs202070685 Chr.17: 43848893-43848893 on Build GRCh38
rs370617554 Chr.17: 43848782-43848782 on Build GRCh38
rs768545849 Chr.17: 43848777-43848777 on Build GRCh38
rs762263543 Chr.17: 43848889-43848889 on Build GRCh38
rs780288721 Chr.17: 43848851-43848851 on Build GRCh38
rs201803597 Chr.17: 43848773-43848773 on Build GRCh38
rs781414083 Chr.17: 43848763-43848763 on Build GRCh38
rs764628224 Chr.17: 43848837-43848837 on Build GRCh38
rs776427136 Chr.17: 43848883-43848883 on Build GRCh38
rs72836561 Chr.17: 43848758-43848758 on Build GRCh38
rs754118437 Chr.17: 43848904-43848904 on Build GRCh38
rs889140352 Chr.17: 43848823-43848823 on Build GRCh38
rs763442534 Chr.17: 43848836-43848836 on Build GRCh38
rs749020643 Chr.17: 43848852-43848852 on Build GRCh38
rs750623950 Chr.17: 43848848-43848848 on Build GRCh38

View Full Product Details