Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 51147370-51147635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf185
Entrez Gene ID
284546
Gene Name
chromosome 1 open reading frame 185
Gene Aliases
-
UniGene
Hs.176177
Gene Chromosome Location
Chr.1: 51102234-51148082 on Build GRCh38
Transcripts
NM_001136508.1 XM_011541282.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3944165 c.459C>G p.D153E Chr.1: 51147630-51147630 on Build GRCh38
4453756 c.343A>T p.I115F Chr.1: 51147514-51147514 on Build GRCh38
4545024 c.368G>A p.S123N Chr.1: 51147539-51147539 on Build GRCh38
4590309 c.341A>G p.N114S Chr.1: 51147512-51147512 on Build GRCh38
1152116 c.360A>G p.S120S Chr.1: 51147531-51147531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531194862 Chr.1: 51147582-51147582 on Build GRCh38
rs910625920 Chr.1: 51147441-51147441 on Build GRCh38
rs754836275 Chr.1: 51147479-51147479 on Build GRCh38
rs764659090 Chr.1: 51147537-51147537 on Build GRCh38
rs1023543760 Chr.1: 51147507-51147507 on Build GRCh38
rs527525254 Chr.1: 51147475-51147475 on Build GRCh38
rs976175779 Chr.1: 51147498-51147498 on Build GRCh38
rs777736480 Chr.1: 51147554-51147554 on Build GRCh38
rs1012127262 Chr.1: 51147503-51147503 on Build GRCh38
rs558092132 Chr.1: 51147488-51147488 on Build GRCh38
rs371914739 Chr.1: 51147539-51147539 on Build GRCh38
rs552051806 Chr.1: 51147512-51147512 on Build GRCh38
rs41286536 Chr.1: 51147540-51147540 on Build GRCh38
rs935829973 Chr.1: 51147563-51147563 on Build GRCh38
rs750808919 Chr.1: 51147482-51147482 on Build GRCh38
rs746071486 Chr.1: 51147583-51147583 on Build GRCh38
rs778656488 Chr.1: 51147496-51147496 on Build GRCh38
rs752614347 Chr.1: 51147497-51147497 on Build GRCh38
rs753636308 Chr.1: 51147476-51147476 on Build GRCh38
rs925923164 Chr.1: 51147553-51147553 on Build GRCh38
rs192084994 Chr.1: 51147411-51147411 on Build GRCh38
rs867196497 Chr.1: 51147487-51147487 on Build GRCh38
rs76431948 Chr.1: 51147573-51147573 on Build GRCh38
rs373551780 Chr.1: 51147473-51147473 on Build GRCh38
rs1053998165 Chr.1: 51147437-51147437 on Build GRCh38

View Full Product Details