Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109689439-109689692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP6
Entrez Gene ID
839
Gene Name
caspase 6
Gene Aliases
MCH2
UniGene
Hs.654616
Gene Chromosome Location
Chr.4: 109688629-109703473 on Build GRCh38
Transcripts
NM_001226.3 NM_032992.2 NR_133012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7336748 c.722C>T p.S241F Chr.4: 109689490-109689490 on Build GRCh38
1049963 c.658C>T p.R220W Chr.4: 109689554-109689554 on Build GRCh38
6992018 c.688C>G p.Q230E Chr.4: 109689524-109689524 on Build GRCh38
6278963 c.672C>T p.N224N Chr.4: 109689540-109689540 on Build GRCh38
6223692 c.644-2A>T p.? Chr.4: 109689570-109689570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753503480 Chr.4: 109689592-109689592 on Build GRCh38
rs910302235 Chr.4: 109689648-109689648 on Build GRCh38
rs528718070 Chr.4: 109689553-109689553 on Build GRCh38
rs573158505 Chr.4: 109689630-109689630 on Build GRCh38
rs139632346 Chr.4: 109689629-109689629 on Build GRCh38
rs770691367 Chr.4: 109689483-109689483 on Build GRCh38
rs756942702 Chr.4: 109689593-109689593 on Build GRCh38
rs777443610 Chr.4: 109689467-109689467 on Build GRCh38
rs944517558 Chr.4: 109689649-109689649 on Build GRCh38
rs750804965 Chr.4: 109689540-109689540 on Build GRCh38
rs140798160 Chr.4: 109689465-109689467 on Build GRCh38
rs768913377 Chr.4: 109689619-109689619 on Build GRCh38
rs747061387 Chr.4: 109689617-109689617 on Build GRCh38
rs749088816 Chr.4: 109689480-109689480 on Build GRCh38
rs181993007 Chr.4: 109689507-109689507 on Build GRCh38
rs752173591 Chr.4: 109689554-109689554 on Build GRCh38
rs745729039 Chr.4: 109689485-109689485 on Build GRCh38
rs369838878 Chr.4: 109689600-109689600 on Build GRCh38
rs575274732 Chr.4: 109689570-109689571 on Build GRCh38
rs771986235 Chr.4: 109689522-109689522 on Build GRCh38
rs951710682 Chr.4: 109689543-109689544 on Build GRCh38
rs775658986 Chr.4: 109689493-109689493 on Build GRCh38
rs374029012 Chr.4: 109689555-109689555 on Build GRCh38
rs961655921 Chr.4: 109689487-109689487 on Build GRCh38
rs968608406 Chr.4: 109689631-109689631 on Build GRCh38
rs765586832 Chr.4: 109689539-109689539 on Build GRCh38
rs774343986 Chr.4: 109689484-109689484 on Build GRCh38
rs777040313 Chr.4: 109689510-109689510 on Build GRCh38
rs772092187 Chr.4: 109689603-109689603 on Build GRCh38
rs758782935 Chr.4: 109689548-109689548 on Build GRCh38
rs376409522 Chr.4: 109689527-109689527 on Build GRCh38
rs373444977 Chr.4: 109689595-109689595 on Build GRCh38
rs772153635 Chr.4: 109689488-109689488 on Build GRCh38
rs764376974 Chr.4: 109689508-109689508 on Build GRCh38
rs777393227 Chr.4: 109689567-109689567 on Build GRCh38
rs755724113 Chr.4: 109689463-109689463 on Build GRCh38
rs780177142 Chr.4: 109689615-109689615 on Build GRCh38

View Full Product Details