Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27056007-27056232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL5
Entrez Gene ID
60509
Gene Name
ATP/GTP binding protein like 5
Gene Aliases
CCP5, RP75
UniGene
Hs.138207
Gene Chromosome Location
Chr.2: 27051423-27070622 on Build GRCh38
Transcripts
NM_001035507.2 NM_021831.5 NR_104246.1 XM_005264477.2 XM_011533011.2 XM_011533012.2 XM_011533013.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182215 c.1274G>A p.S425N Chr.2: 27056047-27056047 on Build GRCh38
5647097 c.1260C>G p.I420M Chr.2: 27056033-27056033 on Build GRCh38
7194030 c.1256_1274del19 p.T419fs*43 Chr.2: 27056029-27056047 on Build GRCh38
5647096 c.1260C>G p.I420M Chr.2: 27056033-27056033 on Build GRCh38
1244638 c.1336G>A p.G446R Chr.2: 27056109-27056109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760778106 Chr.2: 27056108-27056108 on Build GRCh38
rs187389989 Chr.2: 27056037-27056037 on Build GRCh38
rs758590625 Chr.2: 27056179-27056179 on Build GRCh38
rs749076059 Chr.2: 27056167-27056167 on Build GRCh38
rs1050928731 Chr.2: 27056066-27056066 on Build GRCh38
rs775501426 Chr.2: 27056102-27056102 on Build GRCh38
rs186946982 Chr.2: 27056161-27056161 on Build GRCh38
rs757989531 Chr.2: 27056034-27056034 on Build GRCh38
rs773015369 Chr.2: 27056122-27056122 on Build GRCh38
rs769092977 Chr.2: 27056109-27056109 on Build GRCh38
rs377359840 Chr.2: 27056204-27056204 on Build GRCh38
rs762755704 Chr.2: 27056134-27056134 on Build GRCh38
rs747733044 Chr.2: 27056032-27056033 on Build GRCh38
rs752564075 Chr.2: 27056160-27056160 on Build GRCh38
rs912485968 Chr.2: 27056074-27056074 on Build GRCh38
rs746031691 Chr.2: 27056062-27056062 on Build GRCh38
rs932566108 Chr.2: 27056041-27056041 on Build GRCh38
rs191555579 Chr.2: 27056110-27056110 on Build GRCh38
rs1025088650 Chr.2: 27056051-27056051 on Build GRCh38
rs762304587 Chr.2: 27056120-27056120 on Build GRCh38
rs374320938 Chr.2: 27056159-27056159 on Build GRCh38
rs777600997 Chr.2: 27056165-27056165 on Build GRCh38
rs772266754 Chr.2: 27056097-27056097 on Build GRCh38
rs559344816 Chr.2: 27056159-27056160 on Build GRCh38
rs751076244 Chr.2: 27056148-27056148 on Build GRCh38
rs140111286 Chr.2: 27056038-27056038 on Build GRCh38
rs146812730 Chr.2: 27056181-27056181 on Build GRCh38
rs183813669 Chr.2: 27056126-27056126 on Build GRCh38
rs2278593 Chr.2: 27056145-27056145 on Build GRCh38
rs754458465 Chr.2: 27056152-27056152 on Build GRCh38
rs373237623 Chr.2: 27056039-27056039 on Build GRCh38
rs779110735 Chr.2: 27056052-27056052 on Build GRCh38

View Full Product Details