Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169646098-169646367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC173
Entrez Gene ID
129881
Gene Name
coiled-coil domain containing 173
Gene Aliases
C2orf77
UniGene
Hs.370111
Gene Chromosome Location
Chr.2: 169645425-169694421 on Build GRCh38
Transcripts
NM_001085447.1 XM_011510589.1 XM_011510590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5441090 c.1370G>A p.C457Y Chr.2: 169646130-169646130 on Build GRCh38
5928249 c.1402G>A p.E468K Chr.2: 169646098-169646098 on Build GRCh38
6154806 c.1395T>A p.A465A Chr.2: 169646105-169646105 on Build GRCh38
5819430 c.1372A>T p.R458* Chr.2: 169646128-169646128 on Build GRCh38
4526544 c.1401G>A p.K467K Chr.2: 169646099-169646099 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs550534621 Chr.2: 169646216-169646216 on Build GRCh38
rs750178025 Chr.2: 169646141-169646141 on Build GRCh38
rs776281377 Chr.2: 169646191-169646191 on Build GRCh38
rs1042489587 Chr.2: 169646155-169646155 on Build GRCh38
rs530914711 Chr.2: 169646165-169646165 on Build GRCh38
rs62171332 Chr.2: 169646316-169646316 on Build GRCh38
rs768391958 Chr.2: 169646134-169646134 on Build GRCh38
rs993684132 Chr.2: 169646344-169646344 on Build GRCh38
rs1008256857 Chr.2: 169646296-169646296 on Build GRCh38
rs200585776 Chr.2: 169646163-169646163 on Build GRCh38
rs889355066 Chr.2: 169646266-169646266 on Build GRCh38
rs146736463 Chr.2: 169646135-169646135 on Build GRCh38
rs562174057 Chr.2: 169646137-169646137 on Build GRCh38
rs539483202 Chr.2: 169646261-169646261 on Build GRCh38
rs761468830 Chr.2: 169646200-169646200 on Build GRCh38
rs775461717 Chr.2: 169646205-169646209 on Build GRCh38
rs570529136 Chr.2: 169646227-169646227 on Build GRCh38
rs779944728 Chr.2: 169646321-169646321 on Build GRCh38
rs746791084 Chr.2: 169646210-169646215 on Build GRCh38
rs774599656 Chr.2: 169646156-169646156 on Build GRCh38
rs761089406 Chr.2: 169646173-169646173 on Build GRCh38
rs760084395 Chr.2: 169646158-169646158 on Build GRCh38
rs766326087 Chr.2: 169646232-169646232 on Build GRCh38
rs764435052 Chr.2: 169646180-169646180 on Build GRCh38
rs140349520 Chr.2: 169646265-169646265 on Build GRCh38

View Full Product Details