Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92319409-92319650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKIB1
Entrez Gene ID
54467
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
-
UniGene
Hs.83293
Gene Chromosome Location
Chr.7: 92246216-92401773 on Build GRCh38
Transcripts
NM_019004.1 XM_005250458.3 XM_011516334.1 XM_011516335.2 XM_017012359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6341314 c.591G>T p.L197L Chr.7: 92319434-92319434 on Build GRCh38
2864105 c.624G>C p.E208D Chr.7: 92319467-92319467 on Build GRCh38
4988527 c.665G>A p.R222Q Chr.7: 92319508-92319508 on Build GRCh38
318686 c.622G>T p.E208* Chr.7: 92319465-92319465 on Build GRCh38
1187356 c.603G>A p.M201I Chr.7: 92319446-92319446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767516506 Chr.7: 92319550-92319550 on Build GRCh38
rs375124519 Chr.7: 92319464-92319464 on Build GRCh38
rs267601627 Chr.7: 92319463-92319463 on Build GRCh38
rs368340575 Chr.7: 92319497-92319497 on Build GRCh38
rs767687281 Chr.7: 92319478-92319479 on Build GRCh38
rs1047383222 Chr.7: 92319609-92319609 on Build GRCh38
rs773534695 Chr.7: 92319575-92319575 on Build GRCh38
rs200616871 Chr.7: 92319537-92319537 on Build GRCh38
rs377465174 Chr.7: 92319612-92319614 on Build GRCh38
rs761679571 Chr.7: 92319508-92319508 on Build GRCh38
rs191723687 Chr.7: 92319456-92319456 on Build GRCh38
rs753018317 Chr.7: 92319430-92319430 on Build GRCh38
rs1018187376 Chr.7: 92319605-92319605 on Build GRCh38
rs1038519692 Chr.7: 92319553-92319553 on Build GRCh38
rs1056610041 Chr.7: 92319466-92319466 on Build GRCh38
rs754332725 Chr.7: 92319454-92319454 on Build GRCh38
rs535305764 Chr.7: 92319601-92319601 on Build GRCh38
rs756499842 Chr.7: 92319431-92319431 on Build GRCh38
rs771519055 Chr.7: 92319465-92319465 on Build GRCh38
rs183861480 Chr.7: 92319611-92319611 on Build GRCh38
rs779350747 Chr.7: 92319459-92319459 on Build GRCh38
rs976680803 Chr.7: 92319618-92319618 on Build GRCh38
rs779731136 Chr.7: 92319495-92319495 on Build GRCh38
rs756294301 Chr.7: 92319536-92319537 on Build GRCh38
rs201873179 Chr.7: 92319613-92319615 on Build GRCh38
rs371866651 Chr.7: 92319507-92319507 on Build GRCh38
rs778314909 Chr.7: 92319447-92319447 on Build GRCh38
rs769757507 Chr.7: 92319528-92319528 on Build GRCh38
rs144102934 Chr.7: 92319607-92319609 on Build GRCh38
rs773089398 Chr.7: 92319531-92319531 on Build GRCh38
rs750456947 Chr.7: 92319512-92319513 on Build GRCh38

View Full Product Details