Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58352366-58352595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A1BG
Entrez Gene ID
1
Gene Name
alpha-1-B glycoprotein
Gene Aliases
A1B, ABG, GAB, HYST2477
UniGene
Hs.529161
Gene Chromosome Location
Chr.19: 58346806-58353499 on Build GRCh38
Transcripts
NM_130786.3 NR_015380.2

Gene Symbol
A1BG-AS1
Entrez Gene ID
503538
Gene Name
A1BG antisense RNA 1
Gene Aliases
A1BG-AS, A1BGAS, NCRNA00181
UniGene
Hs.709582
Gene Chromosome Location
Chr.19: 58351970-58355183 on Build GRCh38
Transcripts
NM_130786.3 NR_015380.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956658 c.455G>T p.R152L Chr.19: 58352441-58352441 on Build GRCh38
5134981 c.365C>T p.S122L Chr.19: 58352531-58352531 on Build GRCh38
3971321 c.525T>C p.H175H Chr.19: 58352371-58352371 on Build GRCh38
1397306 c.393delC p.G132fs*2 Chr.19: 58352503-58352503 on Build GRCh38
1612999 c.466C>A p.H156N Chr.19: 58352430-58352430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770861923 Chr.19: 58352407-58352407 on Build GRCh38
rs780325447 Chr.19: 58352528-58352529 on Build GRCh38
rs755034046 Chr.19: 58352497-58352497 on Build GRCh38
rs372209543 Chr.19: 58352465-58352465 on Build GRCh38
rs762287675 Chr.19: 58352554-58352554 on Build GRCh38
rs759325835 Chr.19: 58352438-58352438 on Build GRCh38
rs946975067 Chr.19: 58352477-58352478 on Build GRCh38
rs775186281 Chr.19: 58352449-58352449 on Build GRCh38
rs770622370 Chr.19: 58352455-58352455 on Build GRCh38
rs750827392 Chr.19: 58352566-58352566 on Build GRCh38
rs773052847 Chr.19: 58352398-58352398 on Build GRCh38
rs769410420 Chr.19: 58352441-58352441 on Build GRCh38
rs752023317 Chr.19: 58352462-58352462 on Build GRCh38
rs746922135 Chr.19: 58352502-58352502 on Build GRCh38
rs756328834 Chr.19: 58352488-58352490 on Build GRCh38
rs762470632 Chr.19: 58352550-58352550 on Build GRCh38
rs531751714 Chr.19: 58352478-58352478 on Build GRCh38
rs375666222 Chr.19: 58352466-58352466 on Build GRCh38
rs746767778 Chr.19: 58352404-58352404 on Build GRCh38
rs372876362 Chr.19: 58352504-58352504 on Build GRCh38
rs745490543 Chr.19: 58352529-58352529 on Build GRCh38
rs777706453 Chr.19: 58352500-58352500 on Build GRCh38
rs143368389 Chr.19: 58352503-58352503 on Build GRCh38
rs768058729 Chr.19: 58352552-58352552 on Build GRCh38
rs375725715 Chr.19: 58352531-58352531 on Build GRCh38
rs945350883 Chr.19: 58352518-58352518 on Build GRCh38
rs369642410 Chr.19: 58352494-58352494 on Build GRCh38
rs115704323 Chr.19: 58352387-58352387 on Build GRCh38
rs767846542 Chr.19: 58352563-58352563 on Build GRCh38
rs148407798 Chr.19: 58352477-58352477 on Build GRCh38
rs766539685 Chr.19: 58352570-58352570 on Build GRCh38
rs764616664 Chr.19: 58352456-58352456 on Build GRCh38
rs979909537 Chr.19: 58352542-58352542 on Build GRCh38
rs780077361 Chr.19: 58352491-58352491 on Build GRCh38
rs147168349 Chr.19: 58352530-58352530 on Build GRCh38
rs776353453 Chr.19: 58352434-58352434 on Build GRCh38
rs369917094 Chr.19: 58352567-58352567 on Build GRCh38
rs368738532 Chr.19: 58352442-58352442 on Build GRCh38

View Full Product Details