Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 118846146-118846419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EN1
Entrez Gene ID
2019
Gene Name
engrailed homeobox 1
Gene Aliases
-
UniGene
Hs.271977
Gene Chromosome Location
Chr.2: 118842171-118848183 on Build GRCh38
Transcripts
NM_001426.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6322416 c.754C>A p.L252I Chr.2: 118846414-118846414 on Build GRCh38
5354415 c.858C>T p.S286S Chr.2: 118846310-118846310 on Build GRCh38
6653938 c.771C>T p.N257N Chr.2: 118846397-118846397 on Build GRCh38
5394214 c.843T>A p.Y281* Chr.2: 118846325-118846325 on Build GRCh38
6153593 c.808C>G p.L270V Chr.2: 118846360-118846360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377290281 Chr.2: 118846294-118846294 on Build GRCh38
rs568780926 Chr.2: 118846235-118846235 on Build GRCh38
rs572958274 Chr.2: 118846310-118846310 on Build GRCh38
rs773901760 Chr.2: 118846276-118846276 on Build GRCh38
rs140899919 Chr.2: 118846236-118846236 on Build GRCh38
rs745319108 Chr.2: 118846377-118846377 on Build GRCh38
rs550537662 Chr.2: 118846218-118846218 on Build GRCh38
rs973441142 Chr.2: 118846209-118846209 on Build GRCh38
rs372836352 Chr.2: 118846277-118846277 on Build GRCh38
rs756643647 Chr.2: 118846368-118846368 on Build GRCh38
rs761786635 Chr.2: 118846299-118846299 on Build GRCh38
rs764160256 Chr.2: 118846298-118846298 on Build GRCh38
rs990242803 Chr.2: 118846252-118846253 on Build GRCh38
rs907861667 Chr.2: 118846190-118846190 on Build GRCh38
rs764150550 Chr.2: 118846258-118846268 on Build GRCh38
rs781585814 Chr.2: 118846358-118846358 on Build GRCh38
rs547832298 Chr.2: 118846253-118846253 on Build GRCh38
rs77975317 Chr.2: 118846178-118846178 on Build GRCh38
rs753575308 Chr.2: 118846286-118846287 on Build GRCh38
rs755485466 Chr.2: 118846265-118846265 on Build GRCh38
rs981002688 Chr.2: 118846202-118846202 on Build GRCh38
rs373469424 Chr.2: 118846394-118846394 on Build GRCh38
rs955078846 Chr.2: 118846388-118846388 on Build GRCh38
rs906401807 Chr.2: 118846257-118846257 on Build GRCh38
rs771787747 Chr.2: 118846289-118846289 on Build GRCh38
rs753204936 Chr.2: 118846360-118846360 on Build GRCh38
rs942055735 Chr.2: 118846208-118846208 on Build GRCh38
rs566042334 Chr.2: 118846322-118846322 on Build GRCh38
rs770384749 Chr.2: 118846270-118846270 on Build GRCh38
rs748858784 Chr.2: 118846268-118846268 on Build GRCh38
rs755293277 Chr.2: 118846357-118846357 on Build GRCh38
rs368737948 Chr.2: 118846370-118846370 on Build GRCh38
rs78210121 Chr.2: 118846398-118846398 on Build GRCh38
rs765289549 Chr.2: 118846307-118846307 on Build GRCh38
rs377545740 Chr.2: 118846397-118846397 on Build GRCh38
rs779812846 Chr.2: 118846396-118846396 on Build GRCh38
rs751939892 Chr.2: 118846331-118846331 on Build GRCh38

View Full Product Details