Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 89611394-89611135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD5
Entrez Gene ID
1816
Gene Name
dopamine receptor D5
Gene Aliases
DBDR, DRD1B, DRD1L2
UniGene
Hs.380681
Gene Chromosome Location
Chr.4: 9781634-9784009 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Gene Symbol
SLC2A9
Entrez Gene ID
56606
Gene Name
solute carrier family 2 member 9
Gene Aliases
GLUT9, GLUTX, UAQTL2, URATv1
UniGene
Hs.444612
Gene Chromosome Location
Chr.4: 9771125-10040248 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3607186 c.315T>C p.G105G Chr.4: 9782344-9782344 on Build GRCh38
6649304 c.376G>A p.A126T Chr.4: 9782405-9782405 on Build GRCh38
7427554 c.264T>C p.L88L Chr.4: 9782293-9782293 on Build GRCh38
3607188 c.381C>T p.S127S Chr.4: 9782410-9782410 on Build GRCh38
244148 c.445A>G p.K149E Chr.4: 9782474-9782474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775440942 Chr.4: 9782336-9782336 on Build GRCh38
rs761938234 Chr.4: 9782351-9782351 on Build GRCh38
rs761016838 Chr.4: 9782420-9782420 on Build GRCh38
rs374043983 Chr.4: 9782374-9782374 on Build GRCh38
rs769659017 Chr.4: 9782335-9782335 on Build GRCh38
rs755593107 Chr.4: 9782377-9782377 on Build GRCh38
rs935592815 Chr.4: 9782451-9782451 on Build GRCh38
rs747990243 Chr.4: 9782406-9782406 on Build GRCh38
rs756513171 Chr.4: 9782283-9782283 on Build GRCh38
rs370317852 Chr.4: 9782368-9782368 on Build GRCh38
rs148402761 Chr.4: 9782291-9782291 on Build GRCh38
rs765839800 Chr.4: 9782441-9782441 on Build GRCh38
rs201570391 Chr.4: 9782361-9782361 on Build GRCh38
rs761830697 Chr.4: 9782300-9782300 on Build GRCh38
rs765682568 Chr.4: 9782367-9782367 on Build GRCh38
rs575640830 Chr.4: 9782474-9782474 on Build GRCh38
rs746619432 Chr.4: 9782301-9782301 on Build GRCh38
rs768648199 Chr.4: 9782402-9782402 on Build GRCh38
rs760112914 Chr.4: 9782433-9782433 on Build GRCh38
rs368255650 Chr.4: 9782443-9782443 on Build GRCh38
rs755423087 Chr.4: 9782295-9782295 on Build GRCh38
rs773513412 Chr.4: 9782416-9782416 on Build GRCh38
rs372277553 Chr.4: 9782372-9782372 on Build GRCh38
rs776907638 Chr.4: 9782426-9782426 on Build GRCh38
rs772138219 Chr.4: 9782409-9782409 on Build GRCh38
rs985876615 Chr.4: 9782375-9782375 on Build GRCh38
rs546007578 Chr.4: 9782477-9782477 on Build GRCh38
rs756987933 Chr.4: 9782314-9782314 on Build GRCh38
rs534438513 Chr.4: 9782352-9782352 on Build GRCh38
rs780559451 Chr.4: 9782329-9782329 on Build GRCh38
rs2227841 Chr.4: 9782404-9782404 on Build GRCh38
rs6282 Chr.4: 9782292-9782292 on Build GRCh38
rs76288744 Chr.4: 9782293-9782293 on Build GRCh38
rs113695344 Chr.4: 9782347-9782347 on Build GRCh38
rs868700197 Chr.4: 9782463-9782463 on Build GRCh38
rs773390586 Chr.4: 9782442-9782442 on Build GRCh38
rs760170081 Chr.4: 9782365-9782365 on Build GRCh38
rs975877349 Chr.4: 9782296-9782296 on Build GRCh38
rs753351371 Chr.4: 9782485-9782485 on Build GRCh38
rs184288806 Chr.4: 9782353-9782353 on Build GRCh38
rs2227848 Chr.4: 9782281-9782281 on Build GRCh38
rs771111537 Chr.4: 9782421-9782421 on Build GRCh38
rs750070766 Chr.4: 9782464-9782464 on Build GRCh38
rs2227849 Chr.4: 9782358-9782358 on Build GRCh38
rs763394332 Chr.4: 9782450-9782450 on Build GRCh38
rs147038465 Chr.4: 9782330-9782330 on Build GRCh38
rs749266929 Chr.4: 9782338-9782338 on Build GRCh38
rs749213890 Chr.4: 9782397-9782397 on Build GRCh38
rs150176923 Chr.4: 9782452-9782452 on Build GRCh38
rs896687934 Chr.4: 9782469-9782469 on Build GRCh38
rs779592809 Chr.4: 9782380-9782380 on Build GRCh38
rs541886645 Chr.4: 9782369-9782369 on Build GRCh38
rs147655139 Chr.4: 9782345-9782345 on Build GRCh38

View Full Product Details