Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47971137-47971398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP3
Entrez Gene ID
80279
Gene Name
CDK5 regulatory subunit associated protein 3
Gene Aliases
C53, HSF-27, IC53, LZAP, MST016, OK/SW-cl.114, PP1553
UniGene
Hs.20157
Gene Chromosome Location
Chr.17: 47967904-47981786 on Build GRCh38
Transcripts
NM_001278197.1 NM_001278198.1 NM_001278216.1 NM_001278217.1 NM_176096.2 XM_011525297.1 XM_017025164.1 XM_017025165.1 XM_017025166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625579 c.18C>T p.H6H Chr.17: 47971373-47971373 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773965355 Chr.17: 47971238-47971238 on Build GRCh38
rs755326900 Chr.17: 47971161-47971161 on Build GRCh38
rs562748313 Chr.17: 47971205-47971205 on Build GRCh38
rs759998802 Chr.17: 47971274-47971274 on Build GRCh38
rs985377393 Chr.17: 47971170-47971170 on Build GRCh38
rs775030732 Chr.17: 47971219-47971219 on Build GRCh38
rs779945877 Chr.17: 47971319-47971319 on Build GRCh38
rs758437896 Chr.17: 47971178-47971178 on Build GRCh38
rs781106583 Chr.17: 47971196-47971196 on Build GRCh38
rs775983199 Chr.17: 47971359-47971359 on Build GRCh38
rs1034942148 Chr.17: 47971224-47971224 on Build GRCh38
rs757265163 Chr.17: 47971308-47971308 on Build GRCh38
rs771659085 Chr.17: 47971331-47971331 on Build GRCh38
rs2905846 Chr.17: 47971226-47971226 on Build GRCh38
rs755896703 Chr.17: 47971316-47971316 on Build GRCh38
rs1020696506 Chr.17: 47971287-47971287 on Build GRCh38
rs758162320 Chr.17: 47971368-47971369 on Build GRCh38
rs751727370 Chr.17: 47971377-47971377 on Build GRCh38
rs371425673 Chr.17: 47971188-47971188 on Build GRCh38
rs765642339 Chr.17: 47971357-47971357 on Build GRCh38
rs749021224 Chr.17: 47971321-47971321 on Build GRCh38
rs766996334 Chr.17: 47971266-47971266 on Build GRCh38
rs768321009 Chr.17: 47971237-47971237 on Build GRCh38
rs888235813 Chr.17: 47971167-47971167 on Build GRCh38
rs760128705 Chr.17: 47971341-47971341 on Build GRCh38
rs761399838 Chr.17: 47971253-47971253 on Build GRCh38
rs1003348556 Chr.17: 47971218-47971218 on Build GRCh38
rs965255229 Chr.17: 47971194-47971194 on Build GRCh38
rs769576797 Chr.17: 47971213-47971213 on Build GRCh38
rs778740801 Chr.17: 47971334-47971334 on Build GRCh38
rs749007762 Chr.17: 47971223-47971223 on Build GRCh38
rs373306672 Chr.17: 47971300-47971300 on Build GRCh38
rs781125008 Chr.17: 47971314-47971314 on Build GRCh38
rs529021215 Chr.17: 47971184-47971184 on Build GRCh38
rs765594750 Chr.17: 47971275-47971275 on Build GRCh38
rs751705294 Chr.17: 47971301-47971301 on Build GRCh38
rs1018338766 Chr.17: 47971350-47971350 on Build GRCh38
rs777306417 Chr.17: 47971338-47971338 on Build GRCh38
rs752973977 Chr.17: 47971277-47971277 on Build GRCh38
rs1010277838 Chr.17: 47971228-47971228 on Build GRCh38
rs763223898 Chr.17: 47971360-47971360 on Build GRCh38
rs747674667 Chr.17: 47971325-47971325 on Build GRCh38
rs560190393 Chr.17: 47971163-47971163 on Build GRCh38
rs768417843 Chr.17: 47971324-47971324 on Build GRCh38
rs777220265 Chr.17: 47971269-47971269 on Build GRCh38
rs745857180 Chr.17: 47971340-47971340 on Build GRCh38
rs745759161 Chr.17: 47971315-47971315 on Build GRCh38
rs191902054 Chr.17: 47971193-47971193 on Build GRCh38
rs752913153 Chr.17: 47971171-47971171 on Build GRCh38
rs947685007 Chr.17: 47971304-47971304 on Build GRCh38
rs777950478 Chr.17: 47971187-47971187 on Build GRCh38
rs907413090 Chr.17: 47971344-47971344 on Build GRCh38
rs565601221 Chr.17: 47971370-47971370 on Build GRCh38
rs757352543 Chr.17: 47971379-47971379 on Build GRCh38

View Full Product Details