Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47677728-47678001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRF2
Entrez Gene ID
222611
Gene Name
adhesion G protein-coupled receptor F2
Gene Aliases
GPR111, PGR20, hGPCR35
UniGene
Hs.724860
Gene Chromosome Location
Chr.6: 47656590-47697797 on Build GRCh38
Transcripts
NM_153839.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
227759 c.115+1G>A p.? Chr.6: 47677886-47677886 on Build GRCh38
4687855 c.87C>A p.S29R Chr.6: 47677857-47677857 on Build GRCh38
4456729 c.305C>T p.P102L Chr.6: 47677871-47677871 on Build GRCh38
244748 c.115G>A p.G39S Chr.6: 47677885-47677885 on Build GRCh38
4456728 c.101C>T p.P34L Chr.6: 47677871-47677871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369976762 Chr.6: 47677902-47677902 on Build GRCh38
rs188909499 Chr.6: 47677884-47677884 on Build GRCh38
rs779145205 Chr.6: 47677934-47677934 on Build GRCh38
rs779004494 Chr.6: 47677822-47677822 on Build GRCh38
rs769287903 Chr.6: 47677875-47677875 on Build GRCh38
rs1028968379 Chr.6: 47677972-47677972 on Build GRCh38
rs888816835 Chr.6: 47677910-47677910 on Build GRCh38
rs901892790 Chr.6: 47677941-47677941 on Build GRCh38
rs1032821915 Chr.6: 47677950-47677950 on Build GRCh38
rs192562905 Chr.6: 47677967-47677967 on Build GRCh38
rs866836800 Chr.6: 47677834-47677834 on Build GRCh38
rs977212936 Chr.6: 47677760-47677760 on Build GRCh38
rs566036877 Chr.6: 47677892-47677892 on Build GRCh38
rs368777240 Chr.6: 47677816-47677816 on Build GRCh38
rs1045365220 Chr.6: 47677888-47677888 on Build GRCh38
rs757446187 Chr.6: 47677821-47677821 on Build GRCh38
rs776040265 Chr.6: 47677847-47677847 on Build GRCh38
rs759354863 Chr.6: 47677894-47677894 on Build GRCh38
rs976142191 Chr.6: 47677783-47677783 on Build GRCh38
rs781605320 Chr.6: 47677975-47677975 on Build GRCh38
rs940866256 Chr.6: 47677820-47677820 on Build GRCh38
rs377119961 Chr.6: 47677931-47677931 on Build GRCh38
rs376328329 Chr.6: 47677792-47677792 on Build GRCh38
rs112094923 Chr.6: 47677797-47677797 on Build GRCh38
rs957282050 Chr.6: 47677786-47677786 on Build GRCh38
rs373192598 Chr.6: 47677926-47677926 on Build GRCh38
rs766015296 Chr.6: 47677885-47677885 on Build GRCh38
rs746121609 Chr.6: 47677936-47677936 on Build GRCh38
rs752680936 Chr.6: 47677906-47677906 on Build GRCh38
rs1033348874 Chr.6: 47677762-47677762 on Build GRCh38
rs57588858 Chr.6: 47677807-47677807 on Build GRCh38
rs570080121 Chr.6: 47677943-47677943 on Build GRCh38
rs542532789 Chr.6: 47677790-47677790 on Build GRCh38
rs376969688 Chr.6: 47677871-47677871 on Build GRCh38
rs918371449 Chr.6: 47677865-47677865 on Build GRCh38
rs41273680 Chr.6: 47677819-47677819 on Build GRCh38
rs1008563680 Chr.6: 47677918-47677918 on Build GRCh38
rs114015790 Chr.6: 47677937-47677937 on Build GRCh38
rs746199530 Chr.6: 47677833-47677833 on Build GRCh38
rs756180961 Chr.6: 47677907-47677907 on Build GRCh38
rs371960151 Chr.6: 47677803-47677803 on Build GRCh38
rs764143905 Chr.6: 47677908-47677908 on Build GRCh38
rs772766494 Chr.6: 47677880-47677880 on Build GRCh38
rs892987675 Chr.6: 47677749-47677749 on Build GRCh38
rs966030918 Chr.6: 47677777-47677777 on Build GRCh38
rs772342906 Chr.6: 47677842-47677842 on Build GRCh38
rs1020403191 Chr.6: 47677772-47677772 on Build GRCh38
rs1041814038 Chr.6: 47677940-47677940 on Build GRCh38

View Full Product Details