Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200873758-200873983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR25
Entrez Gene ID
2848
Gene Name
G protein-coupled receptor 25
Gene Aliases
-
UniGene
Hs.534316
Gene Chromosome Location
Chr.1: 200872955-200874178 on Build GRCh38
Transcripts
NM_005298.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4688134 c.864C>A p.G288G Chr.1: 200873901-200873901 on Build GRCh38
2211617 c.837C>A p.C279* Chr.1: 200873874-200873874 on Build GRCh38
6674064 c.730C>T p.R244C Chr.1: 200873767-200873767 on Build GRCh38
4688133 c.742G>A p.A248T Chr.1: 200873779-200873779 on Build GRCh38
6674063 c.887C>T p.A296V Chr.1: 200873924-200873924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776242726 Chr.1: 200873827-200873827 on Build GRCh38
rs987098227 Chr.1: 200873961-200873961 on Build GRCh38
rs761919990 Chr.1: 200873912-200873912 on Build GRCh38
rs776086326 Chr.1: 200873885-200873885 on Build GRCh38
rs764570812 Chr.1: 200873841-200873841 on Build GRCh38
rs780727402 Chr.1: 200873876-200873876 on Build GRCh38
rs750312582 Chr.1: 200873838-200873838 on Build GRCh38
rs139739808 Chr.1: 200873846-200873846 on Build GRCh38
rs762164228 Chr.1: 200873845-200873845 on Build GRCh38
rs753717394 Chr.1: 200873777-200873777 on Build GRCh38
rs752625589 Chr.1: 200873932-200873932 on Build GRCh38
rs146592622 Chr.1: 200873859-200873859 on Build GRCh38
rs774898574 Chr.1: 200873888-200873888 on Build GRCh38
rs370410174 Chr.1: 200873858-200873858 on Build GRCh38
rs867237928 Chr.1: 200873897-200873897 on Build GRCh38
rs766503021 Chr.1: 200873928-200873928 on Build GRCh38
rs769140415 Chr.1: 200873887-200873887 on Build GRCh38
rs777349399 Chr.1: 200873803-200873803 on Build GRCh38
rs907085858 Chr.1: 200873789-200873789 on Build GRCh38
rs763346341 Chr.1: 200873840-200873840 on Build GRCh38
rs767732388 Chr.1: 200873915-200873915 on Build GRCh38
rs769428831 Chr.1: 200873883-200873883 on Build GRCh38
rs754985945 Chr.1: 200873784-200873784 on Build GRCh38
rs954505725 Chr.1: 200873895-200873895 on Build GRCh38
rs760742882 Chr.1: 200873925-200873925 on Build GRCh38
rs757029782 Chr.1: 200873964-200873964 on Build GRCh38
rs953930466 Chr.1: 200873959-200873959 on Build GRCh38
rs773337353 Chr.1: 200873919-200873919 on Build GRCh38
rs746721311 Chr.1: 200873804-200873804 on Build GRCh38
rs750566583 Chr.1: 200873847-200873847 on Build GRCh38
rs892790384 Chr.1: 200873863-200873863 on Build GRCh38
rs746335468 Chr.1: 200873829-200873829 on Build GRCh38
rs1011230496 Chr.1: 200873880-200873880 on Build GRCh38
rs777611049 Chr.1: 200873865-200873865 on Build GRCh38
rs1028025618 Chr.1: 200873886-200873886 on Build GRCh38
rs758369263 Chr.1: 200873939-200873939 on Build GRCh38
rs758221692 Chr.1: 200873801-200873801 on Build GRCh38
rs747890063 Chr.1: 200873792-200873792 on Build GRCh38
rs746609301 Chr.1: 200873866-200873866 on Build GRCh38
rs758174965 Chr.1: 200873862-200873862 on Build GRCh38
rs756171622 Chr.1: 200873848-200873848 on Build GRCh38
rs756981429 Chr.1: 200873874-200873874 on Build GRCh38
rs770341765 Chr.1: 200873818-200873818 on Build GRCh38
rs146887111 Chr.1: 200873844-200873844 on Build GRCh38
rs751316948 Chr.1: 200873948-200873948 on Build GRCh38
rs200739616 Chr.1: 200873942-200873942 on Build GRCh38

View Full Product Details