Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81755347-81755580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP4C
Entrez Gene ID
92421
Gene Name
charged multivesicular body protein 4C
Gene Aliases
SNF7-3, Shax3, VPS32C
UniGene
Hs.183861
Gene Chromosome Location
Chr.8: 81732453-81759515 on Build GRCh38
Transcripts
NM_152284.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6193728 c.412C>T p.Q138* Chr.8: 81755413-81755413 on Build GRCh38
5414539 c.423C>T p.I141I Chr.8: 81755424-81755424 on Build GRCh38
1755806 c.400G>C p.E134Q Chr.8: 81755401-81755401 on Build GRCh38
1190805 c.474_475delCT p.F159fs*1 Chr.8: 81755475-81755476 on Build GRCh38
3942789 c.409G>A p.E137K Chr.8: 81755410-81755410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765528700 Chr.8: 81755511-81755511 on Build GRCh38
rs923860424 Chr.8: 81755522-81755522 on Build GRCh38
rs1053732013 Chr.8: 81755550-81755550 on Build GRCh38
rs76125135 Chr.8: 81755498-81755498 on Build GRCh38
rs896068361 Chr.8: 81755546-81755546 on Build GRCh38
rs143113522 Chr.8: 81755455-81755455 on Build GRCh38
rs773648472 Chr.8: 81755514-81755514 on Build GRCh38
rs759677359 Chr.8: 81755526-81755526 on Build GRCh38
rs767124462 Chr.8: 81755395-81755395 on Build GRCh38
rs375563916 Chr.8: 81755484-81755484 on Build GRCh38
rs752058231 Chr.8: 81755519-81755519 on Build GRCh38
rs748814193 Chr.8: 81755409-81755410 on Build GRCh38
rs770651537 Chr.8: 81755525-81755525 on Build GRCh38
rs374378714 Chr.8: 81755518-81755518 on Build GRCh38
rs760326588 Chr.8: 81755505-81755505 on Build GRCh38
rs953790425 Chr.8: 81755459-81755459 on Build GRCh38
rs775170664 Chr.8: 81755401-81755401 on Build GRCh38
rs114287276 Chr.8: 81755456-81755456 on Build GRCh38
rs145626999 Chr.8: 81755488-81755488 on Build GRCh38
rs762154389 Chr.8: 81755420-81755420 on Build GRCh38
rs1015669530 Chr.8: 81755379-81755379 on Build GRCh38
rs370069091 Chr.8: 81755424-81755424 on Build GRCh38
rs767027802 Chr.8: 81755504-81755504 on Build GRCh38
rs780576088 Chr.8: 81755547-81755547 on Build GRCh38
rs766325214 Chr.8: 81755453-81755453 on Build GRCh38
rs953333952 Chr.8: 81755452-81755452 on Build GRCh38
rs368570271 Chr.8: 81755490-81755490 on Build GRCh38
rs755903730 Chr.8: 81755470-81755470 on Build GRCh38
rs556258220 Chr.8: 81755489-81755489 on Build GRCh38
rs528902989 Chr.8: 81755425-81755425 on Build GRCh38
rs758957840 Chr.8: 81755437-81755437 on Build GRCh38
rs774193888 Chr.8: 81755495-81755495 on Build GRCh38
rs771626812 Chr.8: 81755500-81755500 on Build GRCh38
rs960998368 Chr.8: 81755419-81755419 on Build GRCh38

View Full Product Details