Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52464982-52465243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHC1
Entrez Gene ID
114327
Gene Name
EF-hand domain containing 1
Gene Aliases
EJM1, dJ304B14.2
UniGene
Hs.403171
Gene Chromosome Location
Chr.6: 52420196-52495785 on Build GRCh38
Transcripts
NM_001172420.1 NM_018100.3 NR_033327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3355095 c.1114C>T p.R372W Chr.6: 52465092-52465092 on Build GRCh38
6652023 c.1071G>T p.E357D Chr.6: 52465049-52465049 on Build GRCh38
5964585 c.1109G>C p.S370T Chr.6: 52465087-52465087 on Build GRCh38
6652029 c.1102G>T p.D368Y Chr.6: 52465080-52465080 on Build GRCh38
4680325 c.1025T>C p.F342S Chr.6: 52465003-52465003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs505760 Chr.6: 52465047-52465047 on Build GRCh38
rs947160019 Chr.6: 52465075-52465075 on Build GRCh38
rs533695444 Chr.6: 52465039-52465039 on Build GRCh38
rs953235641 Chr.6: 52465008-52465008 on Build GRCh38
rs755819122 Chr.6: 52465142-52465142 on Build GRCh38
rs757987501 Chr.6: 52465088-52465088 on Build GRCh38
rs761135063 Chr.6: 52465051-52465051 on Build GRCh38
rs771054427 Chr.6: 52465158-52465158 on Build GRCh38
rs143751828 Chr.6: 52465068-52465068 on Build GRCh38
rs769745156 Chr.6: 52465033-52465033 on Build GRCh38
rs749162830 Chr.6: 52465151-52465151 on Build GRCh38
rs767833659 Chr.6: 52465042-52465042 on Build GRCh38
rs1051652198 Chr.6: 52465186-52465186 on Build GRCh38
rs377732929 Chr.6: 52465036-52465036 on Build GRCh38
rs549508905 Chr.6: 52465101-52465101 on Build GRCh38
rs535905126 Chr.6: 52465093-52465093 on Build GRCh38
rs201118016 Chr.6: 52465032-52465032 on Build GRCh38
rs527295360 Chr.6: 52465035-52465035 on Build GRCh38
rs754355317 Chr.6: 52465082-52465082 on Build GRCh38
rs760228198 Chr.6: 52465130-52465132 on Build GRCh38
rs780892940 Chr.6: 52465125-52465125 on Build GRCh38
rs781028408 Chr.6: 52465022-52465022 on Build GRCh38
rs115160207 Chr.6: 52465146-52465146 on Build GRCh38
rs200024100 Chr.6: 52465037-52465037 on Build GRCh38
rs1055966349 Chr.6: 52465202-52465202 on Build GRCh38
rs534707843 Chr.6: 52465183-52465183 on Build GRCh38
rs890394671 Chr.6: 52465197-52465197 on Build GRCh38
rs374198314 Chr.6: 52465203-52465203 on Build GRCh38
rs573582894 Chr.6: 52465074-52465074 on Build GRCh38
rs937533552 Chr.6: 52465159-52465159 on Build GRCh38
rs929940195 Chr.6: 52465136-52465136 on Build GRCh38
rs994812402 Chr.6: 52465031-52465031 on Build GRCh38
rs371151471 Chr.6: 52465092-52465092 on Build GRCh38
rs747957348 Chr.6: 52465130-52465130 on Build GRCh38
rs915630036 Chr.6: 52465071-52465071 on Build GRCh38

View Full Product Details