Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41272037-41272292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6031566 c.413delG p.Y140fs*79 Chr.19: 41272205-41272205 on Build GRCh38
4952536 c.551T>G p.F184C Chr.19: 41272214-41272214 on Build GRCh38
4952537 c.422T>G p.F141C Chr.19: 41272214-41272214 on Build GRCh38
3378752 c.520C>T p.P174S Chr.19: 41272183-41272183 on Build GRCh38
4952573 c.565G>T p.D189Y Chr.19: 41272228-41272228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750508404 Chr.19: 41272105-41272105 on Build GRCh38
rs777007765 Chr.19: 41272058-41272058 on Build GRCh38
rs916062948 Chr.19: 41272261-41272261 on Build GRCh38
rs755338186 Chr.19: 41272131-41272131 on Build GRCh38
rs147857804 Chr.19: 41272232-41272232 on Build GRCh38
rs147614451 Chr.19: 41272095-41272095 on Build GRCh38
rs762094204 Chr.19: 41272209-41272209 on Build GRCh38
rs767840371 Chr.19: 41272260-41272260 on Build GRCh38
rs1009197974 Chr.19: 41272164-41272164 on Build GRCh38
rs972056265 Chr.19: 41272091-41272091 on Build GRCh38
rs766813390 Chr.19: 41272109-41272109 on Build GRCh38
rs886390648 Chr.19: 41272163-41272163 on Build GRCh38
rs140550135 Chr.19: 41272199-41272199 on Build GRCh38
rs778467769 Chr.19: 41272188-41272188 on Build GRCh38
rs868518443 Chr.19: 41272184-41272184 on Build GRCh38
rs964879463 Chr.19: 41272187-41272187 on Build GRCh38
rs574973882 Chr.19: 41272248-41272248 on Build GRCh38
rs376250609 Chr.19: 41272172-41272172 on Build GRCh38
rs756946285 Chr.19: 41272183-41272183 on Build GRCh38
rs144502015 Chr.19: 41272205-41272205 on Build GRCh38
rs866322796 Chr.19: 41272056-41272056 on Build GRCh38
rs994528251 Chr.19: 41272096-41272096 on Build GRCh38
rs751907931 Chr.19: 41272128-41272128 on Build GRCh38
rs762971005 Chr.19: 41272223-41272223 on Build GRCh38
rs770189529 Chr.19: 41272215-41272215 on Build GRCh38
rs777346071 Chr.19: 41272142-41272142 on Build GRCh38
rs776633686 Chr.19: 41272207-41272207 on Build GRCh38
rs774732052 Chr.19: 41272239-41272239 on Build GRCh38
rs763494208 Chr.19: 41272108-41272108 on Build GRCh38
rs773293263 Chr.19: 41272221-41272221 on Build GRCh38
rs746763454 Chr.19: 41272204-41272204 on Build GRCh38
rs772007874 Chr.19: 41272198-41272198 on Build GRCh38
rs762182248 Chr.19: 41272065-41272065 on Build GRCh38
rs201362715 Chr.19: 41272165-41272165 on Build GRCh38
rs200908886 Chr.19: 41272197-41272197 on Build GRCh38

View Full Product Details