Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152962816-152963083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHDC1
Entrez Gene ID
85462
Gene Name
FH2 domain containing 1
Gene Aliases
-
UniGene
Hs.132629
Gene Chromosome Location
Chr.4: 152911570-152979696 on Build GRCh38
Transcripts
NM_033393.2 XM_005263319.3 XM_006714393.3 XM_011532388.1 XM_011532389.1 XM_017008787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285043 c.900G>A p.Q300Q Chr.4: 152962863-152962863 on Build GRCh38
7408979 c.925G>T p.G309W Chr.4: 152963026-152963026 on Build GRCh38
5865889 c.943G>A p.V315I Chr.4: 152963044-152963044 on Build GRCh38
7078048 c.934G>A p.G312S Chr.4: 152963035-152963035 on Build GRCh38
5865890 c.962C>T p.S321F Chr.4: 152963063-152963063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778220243 Chr.4: 152962951-152962952 on Build GRCh38
rs568875801 Chr.4: 152962953-152962954 on Build GRCh38
rs879876978 Chr.4: 152962945-152962945 on Build GRCh38
rs550609626 Chr.4: 152962953-152962953 on Build GRCh38
rs747507922 Chr.4: 152963027-152963027 on Build GRCh38
rs529081495 Chr.4: 152962947-152962953 on Build GRCh38
rs763712321 Chr.4: 152962979-152962980 on Build GRCh38
rs769014919 Chr.4: 152962918-152962918 on Build GRCh38
rs72725633 Chr.4: 152962945-152962945 on Build GRCh38
rs867920226 Chr.4: 152962968-152962968 on Build GRCh38
rs150771735 Chr.4: 152962862-152962862 on Build GRCh38
rs561915155 Chr.4: 152962945-152962951 on Build GRCh38
rs773637282 Chr.4: 152962987-152962987 on Build GRCh38
rs775453538 Chr.4: 152962990-152962990 on Build GRCh38
rs764869341 Chr.4: 152962980-152962983 on Build GRCh38
rs774555201 Chr.4: 152962902-152962902 on Build GRCh38
rs58275172 Chr.4: 152962982-152962983 on Build GRCh38
rs187820141 Chr.4: 152963012-152963012 on Build GRCh38
rs561509868 Chr.4: 152962942-152962942 on Build GRCh38
rs200235057 Chr.4: 152963034-152963034 on Build GRCh38
rs951160478 Chr.4: 152962955-152962955 on Build GRCh38
rs372962979 Chr.4: 152963002-152963002 on Build GRCh38
rs34181980 Chr.4: 152962981-152962982 on Build GRCh38
rs555361313 Chr.4: 152962944-152962945 on Build GRCh38
rs760565225 Chr.4: 152962991-152962991 on Build GRCh38
rs377643778 Chr.4: 152962940-152962941 on Build GRCh38
rs1009270136 Chr.4: 152962977-152962978 on Build GRCh38
rs757908286 Chr.4: 152962882-152962882 on Build GRCh38
rs756911459 Chr.4: 152962868-152962868 on Build GRCh38
rs763057872 Chr.4: 152962988-152962988 on Build GRCh38
rs773547122 Chr.4: 152963057-152963057 on Build GRCh38
rs778333848 Chr.4: 152962874-152962874 on Build GRCh38
rs753649521 Chr.4: 152963001-152963001 on Build GRCh38
rs763811831 Chr.4: 152962994-152962994 on Build GRCh38
rs779580537 Chr.4: 152962885-152962886 on Build GRCh38
rs756340958 Chr.4: 152962993-152962994 on Build GRCh38
rs369698283 Chr.4: 152962915-152962915 on Build GRCh38
rs1033713104 Chr.4: 152962947-152962947 on Build GRCh38
rs879146364 Chr.4: 152962948-152962949 on Build GRCh38
rs1025004147 Chr.4: 152962892-152962892 on Build GRCh38
rs745368122 Chr.4: 152962875-152962875 on Build GRCh38
rs397995844 Chr.4: 152962981-152962982 on Build GRCh38
rs777900441 Chr.4: 152963030-152963031 on Build GRCh38
rs748728112 Chr.4: 152962947-152962948 on Build GRCh38
rs532929055 Chr.4: 152962991-152962991 on Build GRCh38
rs749498428 Chr.4: 152963059-152963059 on Build GRCh38
rs892751106 Chr.4: 152963060-152963060 on Build GRCh38
rs766419291 Chr.4: 152962989-152962989 on Build GRCh38
rs74812681 Chr.4: 152962983-152962983 on Build GRCh38
rs370344776 Chr.4: 152963052-152963052 on Build GRCh38
rs760371971 Chr.4: 152962978-152962979 on Build GRCh38
rs906480728 Chr.4: 152963021-152963021 on Build GRCh38
rs764862826 Chr.4: 152963007-152963007 on Build GRCh38
rs879130492 Chr.4: 152962951-152962951 on Build GRCh38
rs767745698 Chr.4: 152962975-152962975 on Build GRCh38
rs750200704 Chr.4: 152963035-152963035 on Build GRCh38
rs878874451 Chr.4: 152962941-152962948 on Build GRCh38
rs763903554 Chr.4: 152962856-152962856 on Build GRCh38
rs777165071 Chr.4: 152962926-152962926 on Build GRCh38
rs757902505 Chr.4: 152963011-152963011 on Build GRCh38
rs780574762 Chr.4: 152962894-152962894 on Build GRCh38
rs972766921 Chr.4: 152963031-152963031 on Build GRCh38
rs879204084 Chr.4: 152962942-152962943 on Build GRCh38
rs796312794 Chr.4: 152962953-152962953 on Build GRCh38
rs748520934 Chr.4: 152963045-152963045 on Build GRCh38
rs112231596 Chr.4: 152962849-152962849 on Build GRCh38
rs750079434 Chr.4: 152963008-152963008 on Build GRCh38
rs963723845 Chr.4: 152962949-152962949 on Build GRCh38
rs775269266 Chr.4: 152962975-152962976 on Build GRCh38
rs200601461 Chr.4: 152962979-152962979 on Build GRCh38
rs796264890 Chr.4: 152962982-152962983 on Build GRCh38

View Full Product Details