Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31970715-31970960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DXO
Entrez Gene ID
1797
Gene Name
decapping exoribonuclease
Gene Aliases
DOM3L, DOM3Z, NG6, RAI1
UniGene
Hs.153299
Gene Chromosome Location
Chr.6: 31969811-31972290 on Build GRCh38
Transcripts
NM_005510.3 XM_006715005.3 XM_017010329.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
329664 c.696_697delAG p.E234fs*61 Chr.6: 31970721-31970722 on Build GRCh38
3830157 c.618C>T p.P206P Chr.6: 31970800-31970800 on Build GRCh38
260563 c.593-9C>A p.? Chr.6: 31970834-31970834 on Build GRCh38
4654090 c.546G>A p.R182R Chr.6: 31970958-31970958 on Build GRCh38
3861929 c.544C>T p.R182W Chr.6: 31970960-31970960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755484544 Chr.6: 31970791-31970791 on Build GRCh38
rs758713594 Chr.6: 31970911-31970911 on Build GRCh38
rs1041528285 Chr.6: 31970901-31970901 on Build GRCh38
rs894065801 Chr.6: 31970870-31970870 on Build GRCh38
rs1019441981 Chr.6: 31970932-31970932 on Build GRCh38
rs947520385 Chr.6: 31970887-31970887 on Build GRCh38
rs747204153 Chr.6: 31970913-31970913 on Build GRCh38
rs776629971 Chr.6: 31970834-31970834 on Build GRCh38
rs764008054 Chr.6: 31970851-31970851 on Build GRCh38
rs943728894 Chr.6: 31970907-31970907 on Build GRCh38
rs762895946 Chr.6: 31970836-31970836 on Build GRCh38
rs567817660 Chr.6: 31970750-31970750 on Build GRCh38
rs758044504 Chr.6: 31970920-31970921 on Build GRCh38
rs747205691 Chr.6: 31970824-31970824 on Build GRCh38
rs778475378 Chr.6: 31970874-31970879 on Build GRCh38
rs796615211 Chr.6: 31970785-31970785 on Build GRCh38
rs914773536 Chr.6: 31970873-31970873 on Build GRCh38
rs144960609 Chr.6: 31970812-31970812 on Build GRCh38
rs752118229 Chr.6: 31970764-31970765 on Build GRCh38
rs771031921 Chr.6: 31970832-31970832 on Build GRCh38
rs749997061 Chr.6: 31970756-31970756 on Build GRCh38
rs767870949 Chr.6: 31970849-31970850 on Build GRCh38
rs781310223 Chr.6: 31970927-31970927 on Build GRCh38
rs773286220 Chr.6: 31970820-31970820 on Build GRCh38
rs755597076 Chr.6: 31970757-31970757 on Build GRCh38
rs754678526 Chr.6: 31970856-31970857 on Build GRCh38
rs548171157 Chr.6: 31970917-31970917 on Build GRCh38
rs753389054 Chr.6: 31970855-31970856 on Build GRCh38
rs749816912 Chr.6: 31970779-31970779 on Build GRCh38
rs200954579 Chr.6: 31970738-31970738 on Build GRCh38
rs748455311 Chr.6: 31970795-31970795 on Build GRCh38
rs767044016 Chr.6: 31970751-31970751 on Build GRCh38
rs199970782 Chr.6: 31970804-31970804 on Build GRCh38
rs755606792 Chr.6: 31970881-31970881 on Build GRCh38
rs779667539 Chr.6: 31970773-31970773 on Build GRCh38
rs750071146 Chr.6: 31970856-31970856 on Build GRCh38
rs41267114 Chr.6: 31970854-31970854 on Build GRCh38
rs763885299 Chr.6: 31970737-31970737 on Build GRCh38
rs535219642 Chr.6: 31970853-31970853 on Build GRCh38
rs745947009 Chr.6: 31970936-31970936 on Build GRCh38
rs899467834 Chr.6: 31970912-31970912 on Build GRCh38
rs752216181 Chr.6: 31970873-31970874 on Build GRCh38
rs766000650 Chr.6: 31970861-31970861 on Build GRCh38
rs416002 Chr.6: 31970858-31970858 on Build GRCh38
rs779530019 Chr.6: 31970892-31970892 on Build GRCh38
rs762105628 Chr.6: 31970806-31970807 on Build GRCh38
rs753156292 Chr.6: 31970899-31970899 on Build GRCh38
rs902928306 Chr.6: 31970902-31970902 on Build GRCh38
rs772380423 Chr.6: 31970803-31970803 on Build GRCh38

View Full Product Details