Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61341710-61341981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TKFC
Entrez Gene ID
26007
Gene Name
triokinase and FMN cyclase
Gene Aliases
DAK, NET45
UniGene
Hs.6278
Gene Chromosome Location
Chr.11: 61333182-61353426 on Build GRCh38
Transcripts
NM_015533.3 XM_011544912.2 XM_017017517.1 XM_017017518.1 XM_017017519.1 XM_017017520.1 XM_017017521.1 XM_017017522.1 XM_017017523.1 XM_017017524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604861 c.613C>T p.P205S Chr.11: 61341870-61341870 on Build GRCh38
1492638 c.617C>A p.T206N Chr.11: 61341874-61341874 on Build GRCh38
6639965 c.566-1G>A p.? Chr.11: 61341822-61341822 on Build GRCh38
369146 c.637G>A p.E213K Chr.11: 61341894-61341894 on Build GRCh38
5956177 c.643G>A p.E215K Chr.11: 61341900-61341900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958141083 Chr.11: 61341863-61341863 on Build GRCh38
rs755294751 Chr.11: 61341891-61341891 on Build GRCh38
rs1034005122 Chr.11: 61341787-61341787 on Build GRCh38
rs770783738 Chr.11: 61341808-61341808 on Build GRCh38
rs764735611 Chr.11: 61341934-61341934 on Build GRCh38
rs369635726 Chr.11: 61341848-61341848 on Build GRCh38
rs142253953 Chr.11: 61341828-61341828 on Build GRCh38
rs773995453 Chr.11: 61341820-61341820 on Build GRCh38
rs760193931 Chr.11: 61341832-61341832 on Build GRCh38
rs895547384 Chr.11: 61341739-61341739 on Build GRCh38
rs372731860 Chr.11: 61341895-61341895 on Build GRCh38
rs955494943 Chr.11: 61341854-61341854 on Build GRCh38
rs756264858 Chr.11: 61341912-61341912 on Build GRCh38
rs1048739852 Chr.11: 61341955-61341955 on Build GRCh38
rs181098169 Chr.11: 61341777-61341777 on Build GRCh38
rs751835009 Chr.11: 61341890-61341890 on Build GRCh38
rs759151407 Chr.11: 61341879-61341879 on Build GRCh38
rs745641149 Chr.11: 61341941-61341941 on Build GRCh38
rs999977168 Chr.11: 61341843-61341843 on Build GRCh38
rs143435164 Chr.11: 61341829-61341829 on Build GRCh38
rs777953943 Chr.11: 61341913-61341913 on Build GRCh38
rs149571662 Chr.11: 61341855-61341855 on Build GRCh38
rs749298721 Chr.11: 61341921-61341921 on Build GRCh38
rs781550668 Chr.11: 61341894-61341894 on Build GRCh38
rs773293843 Chr.11: 61341805-61341805 on Build GRCh38
rs374087598 Chr.11: 61341806-61341806 on Build GRCh38
rs778835536 Chr.11: 61341938-61341938 on Build GRCh38
rs543263692 Chr.11: 61341793-61341793 on Build GRCh38
rs895383128 Chr.11: 61341796-61341796 on Build GRCh38
rs769908556 Chr.11: 61341782-61341782 on Build GRCh38
rs1041566073 Chr.11: 61341783-61341783 on Build GRCh38
rs770744317 Chr.11: 61341924-61341924 on Build GRCh38
rs150473332 Chr.11: 61341831-61341831 on Build GRCh38
rs1020654000 Chr.11: 61341937-61341953 on Build GRCh38
rs117064389 Chr.11: 61341833-61341833 on Build GRCh38
rs573143928 Chr.11: 61341952-61341952 on Build GRCh38

View Full Product Details