Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39631351-39631624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEYL
Entrez Gene ID
26508
Gene Name
hes related family bHLH transcription factor with YRPW motif-like
Gene Aliases
HESR3, HEY3, HRT3, bHLHb33
UniGene
Hs.472566
Gene Chromosome Location
Chr.1: 39623431-39639676 on Build GRCh38
Transcripts
NM_014571.3 XM_005270745.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5532297 c.219C>T p.A73A Chr.1: 39631508-39631508 on Build GRCh38
5734766 c.149T>A p.I50N Chr.1: 39631578-39631578 on Build GRCh38
3489482 c.200G>A p.R67Q Chr.1: 39631527-39631527 on Build GRCh38
7424775 c.172C>T p.R58C Chr.1: 39631555-39631555 on Build GRCh38
3805177 c.164G>A p.R55H Chr.1: 39631563-39631563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772415884 Chr.1: 39631470-39631470 on Build GRCh38
rs751498805 Chr.1: 39631528-39631528 on Build GRCh38
rs777598242 Chr.1: 39631560-39631560 on Build GRCh38
rs755774767 Chr.1: 39631557-39631557 on Build GRCh38
rs928796148 Chr.1: 39631394-39631394 on Build GRCh38
rs143373894 Chr.1: 39631524-39631524 on Build GRCh38
rs772502405 Chr.1: 39631564-39631564 on Build GRCh38
rs918891288 Chr.1: 39631539-39631539 on Build GRCh38
rs747706788 Chr.1: 39631555-39631555 on Build GRCh38
rs141413445 Chr.1: 39631554-39631554 on Build GRCh38
rs1020216057 Chr.1: 39631386-39631386 on Build GRCh38
rs201520391 Chr.1: 39631577-39631577 on Build GRCh38
rs776645048 Chr.1: 39631487-39631487 on Build GRCh38
rs145971519 Chr.1: 39631469-39631469 on Build GRCh38
rs139605272 Chr.1: 39631567-39631567 on Build GRCh38
rs760616348 Chr.1: 39631483-39631483 on Build GRCh38
rs769086936 Chr.1: 39631574-39631574 on Build GRCh38
rs111993765 Chr.1: 39631426-39631426 on Build GRCh38
rs188420721 Chr.1: 39631395-39631395 on Build GRCh38
rs772056146 Chr.1: 39631599-39631599 on Build GRCh38
rs1015499817 Chr.1: 39631575-39631575 on Build GRCh38
rs761992572 Chr.1: 39631496-39631496 on Build GRCh38
rs749545276 Chr.1: 39631456-39631456 on Build GRCh38
rs185263340 Chr.1: 39631384-39631384 on Build GRCh38
rs930297425 Chr.1: 39631563-39631563 on Build GRCh38
rs932389013 Chr.1: 39631481-39631481 on Build GRCh38
rs754936352 Chr.1: 39631536-39631536 on Build GRCh38
rs549868767 Chr.1: 39631421-39631421 on Build GRCh38
rs764225869 Chr.1: 39631486-39631486 on Build GRCh38
rs745657046 Chr.1: 39631502-39631502 on Build GRCh38
rs149665255 Chr.1: 39631591-39631591 on Build GRCh38
rs765492005 Chr.1: 39631504-39631504 on Build GRCh38
rs766168978 Chr.1: 39631525-39631525 on Build GRCh38
rs982030010 Chr.1: 39631391-39631391 on Build GRCh38
rs555240790 Chr.1: 39631510-39631510 on Build GRCh38
rs1170446 Chr.1: 39631460-39631460 on Build GRCh38
rs367583397 Chr.1: 39631437-39631437 on Build GRCh38
rs111738259 Chr.1: 39631431-39631431 on Build GRCh38
rs898414530 Chr.1: 39631576-39631576 on Build GRCh38
rs776072942 Chr.1: 39631474-39631474 on Build GRCh38
rs775779178 Chr.1: 39631566-39631566 on Build GRCh38
rs139786829 Chr.1: 39631561-39631561 on Build GRCh38
rs11206504 Chr.1: 39631464-39631464 on Build GRCh38

View Full Product Details