Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 47737459-47737692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPD
Entrez Gene ID
1052
Gene Name
CCAAT/enhancer binding protein delta
Gene Aliases
C/EBP-delta, CELF, CRP3, NF-IL6-beta
UniGene
Hs.440829
Gene Chromosome Location
Chr.8: 47736914-47738164 on Build GRCh38
Transcripts
NM_005195.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1187535 c.532C>T p.R178W Chr.8: 47737589-47737589 on Build GRCh38
1755735 c.607A>G p.I203V Chr.8: 47737514-47737514 on Build GRCh38
5038725 c.641_649delGGCGCAACC p.R214_N216delRRN Chr.8: 47737472-47737480 on Build GRCh38
7314326 c.661C>A p.Q221K Chr.8: 47737460-47737460 on Build GRCh38
6625958 c.618delC p.K207fs*>63 Chr.8: 47737503-47737503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35029833 Chr.8: 47737509-47737509 on Build GRCh38
rs1023496807 Chr.8: 47737594-47737594 on Build GRCh38
rs774169573 Chr.8: 47737545-47737545 on Build GRCh38
rs762648207 Chr.8: 47737536-47737536 on Build GRCh38
rs750673306 Chr.8: 47737574-47737574 on Build GRCh38
rs770705158 Chr.8: 47737537-47737537 on Build GRCh38
rs780541037 Chr.8: 47737579-47737579 on Build GRCh38
rs769172758 Chr.8: 47737609-47737609 on Build GRCh38
rs781551832 Chr.8: 47737602-47737602 on Build GRCh38
rs758822964 Chr.8: 47737494-47737494 on Build GRCh38
rs111900648 Chr.8: 47737503-47737503 on Build GRCh38
rs541685864 Chr.8: 47737514-47737514 on Build GRCh38
rs769421427 Chr.8: 47737512-47737512 on Build GRCh38
rs753893023 Chr.8: 47737554-47737554 on Build GRCh38
rs751992177 Chr.8: 47737588-47737588 on Build GRCh38
rs769334462 Chr.8: 47737501-47737501 on Build GRCh38
rs1017310803 Chr.8: 47737621-47737621 on Build GRCh38
rs370948191 Chr.8: 47737575-47737575 on Build GRCh38
rs772748708 Chr.8: 47737524-47737524 on Build GRCh38
rs767389697 Chr.8: 47737553-47737553 on Build GRCh38
rs962752316 Chr.8: 47737628-47737628 on Build GRCh38
rs200641900 Chr.8: 47737591-47737591 on Build GRCh38
rs371024952 Chr.8: 47737502-47737502 on Build GRCh38
rs578188520 Chr.8: 47737603-47737603 on Build GRCh38
rs746185483 Chr.8: 47737489-47737489 on Build GRCh38
rs973201486 Chr.8: 47737630-47737630 on Build GRCh38
rs777407313 Chr.8: 47737616-47737616 on Build GRCh38
rs759423146 Chr.8: 47737550-47737550 on Build GRCh38
rs747489027 Chr.8: 47737498-47737498 on Build GRCh38
rs780633015 Chr.8: 47737497-47737497 on Build GRCh38
rs918699325 Chr.8: 47737648-47737648 on Build GRCh38
rs545658644 Chr.8: 47737633-47737633 on Build GRCh38
rs779011097 Chr.8: 47737481-47737481 on Build GRCh38
rs761817545 Chr.8: 47737564-47737564 on Build GRCh38
rs867621033 Chr.8: 47737619-47737619 on Build GRCh38

View Full Product Details