Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92266655-92266898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLMN
Entrez Gene ID
11146
Gene Name
glomulin, FKBP associated protein
Gene Aliases
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM
UniGene
Hs.49105
Gene Chromosome Location
Chr.1: 92245786-92441936 on Build GRCh38
Transcripts
NM_001319683.1 NM_053274.2 NR_135089.1 XM_005270401.3 XM_006710309.2 XM_011540546.2 XM_017000137.1 XM_017000138.1 XM_017000139.1 XM_017000140.1 XM_017000141.1 XM_017000142.1 XM_017000143.1 XM_017000144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4409583 c.1120C>T p.L374F Chr.1: 92266720-92266720 on Build GRCh38
3493356 c.1126C>T p.P376S Chr.1: 92266714-92266714 on Build GRCh38
6191864 c.1116G>A p.M372I Chr.1: 92266724-92266724 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777896358 Chr.1: 92266749-92266750 on Build GRCh38
rs911999460 Chr.1: 92266856-92266856 on Build GRCh38
rs1030476348 Chr.1: 92266805-92266805 on Build GRCh38
rs994742702 Chr.1: 92266758-92266758 on Build GRCh38
rs866074360 Chr.1: 92266777-92266777 on Build GRCh38
rs766973740 Chr.1: 92266766-92266766 on Build GRCh38
rs200121766 Chr.1: 92266745-92266745 on Build GRCh38
rs908152657 Chr.1: 92266836-92266836 on Build GRCh38
rs764995587 Chr.1: 92266776-92266776 on Build GRCh38
rs761219121 Chr.1: 92266764-92266764 on Build GRCh38
rs1012622511 Chr.1: 92266691-92266691 on Build GRCh38
rs987584073 Chr.1: 92266837-92266837 on Build GRCh38
rs758962318 Chr.1: 92266771-92266771 on Build GRCh38
rs776247053 Chr.1: 92266770-92266770 on Build GRCh38
rs772497879 Chr.1: 92266715-92266715 on Build GRCh38
rs964764387 Chr.1: 92266869-92266869 on Build GRCh38
rs984116849 Chr.1: 92266834-92266834 on Build GRCh38
rs1026158794 Chr.1: 92266794-92266794 on Build GRCh38
rs376145913 Chr.1: 92266769-92266769 on Build GRCh38
rs759042428 Chr.1: 92266710-92266710 on Build GRCh38
rs774424609 Chr.1: 92266706-92266706 on Build GRCh38
rs997864173 Chr.1: 92266796-92266796 on Build GRCh38
rs954836603 Chr.1: 92266815-92266815 on Build GRCh38

View Full Product Details