Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79730389-79730648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMYA5
Entrez Gene ID
202333
Gene Name
cardiomyopathy associated 5
Gene Aliases
C5orf10, SPRYD2, TRIM76
UniGene
Hs.482625
Gene Chromosome Location
Chr.5: 79689339-79800226 on Build GRCh38
Transcripts
NM_153610.4 XM_017009212.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3856498 c.1681G>T p.E561* Chr.5: 79730446-79730446 on Build GRCh38
6104696 c.1762G>T p.V588F Chr.5: 79730527-79730527 on Build GRCh38
6631041 c.1630G>A p.E544K Chr.5: 79730395-79730395 on Build GRCh38
1201496 c.1766C>A p.S589Y Chr.5: 79730531-79730531 on Build GRCh38
6104695 c.1762G>T p.V588F Chr.5: 79730527-79730527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770228255 Chr.5: 79730566-79730566 on Build GRCh38
rs781515245 Chr.5: 79730556-79730556 on Build GRCh38
rs757742595 Chr.5: 79730452-79730459 on Build GRCh38
rs372336841 Chr.5: 79730473-79730473 on Build GRCh38
rs779181647 Chr.5: 79730485-79730485 on Build GRCh38
rs35928567 Chr.5: 79730611-79730611 on Build GRCh38
rs746478751 Chr.5: 79730544-79730544 on Build GRCh38
rs921455644 Chr.5: 79730454-79730454 on Build GRCh38
rs369818294 Chr.5: 79730449-79730449 on Build GRCh38
rs759953390 Chr.5: 79730464-79730464 on Build GRCh38
rs72635619 Chr.5: 79730547-79730547 on Build GRCh38
rs749019404 Chr.5: 79730409-79730409 on Build GRCh38
rs751583591 Chr.5: 79730478-79730478 on Build GRCh38
rs200387533 Chr.5: 79730516-79730516 on Build GRCh38
rs201429642 Chr.5: 79730502-79730502 on Build GRCh38
rs778556799 Chr.5: 79730509-79730509 on Build GRCh38
rs746275998 Chr.5: 79730509-79730510 on Build GRCh38
rs777102367 Chr.5: 79730462-79730462 on Build GRCh38
rs572599993 Chr.5: 79730427-79730427 on Build GRCh38
rs753816970 Chr.5: 79730570-79730570 on Build GRCh38
rs896420968 Chr.5: 79730524-79730524 on Build GRCh38
rs975565074 Chr.5: 79730439-79730439 on Build GRCh38
rs867952885 Chr.5: 79730572-79730572 on Build GRCh38
rs760720422 Chr.5: 79730487-79730487 on Build GRCh38
rs753725239 Chr.5: 79730607-79730607 on Build GRCh38
rs768745226 Chr.5: 79730582-79730582 on Build GRCh38
rs758800626 Chr.5: 79730545-79730556 on Build GRCh38
rs752231445 Chr.5: 79730438-79730440 on Build GRCh38
rs763330047 Chr.5: 79730580-79730580 on Build GRCh38
rs182125509 Chr.5: 79730587-79730587 on Build GRCh38
rs373188852 Chr.5: 79730596-79730596 on Build GRCh38
rs779471027 Chr.5: 79730489-79730490 on Build GRCh38
rs771329189 Chr.5: 79730458-79730458 on Build GRCh38
rs376402768 Chr.5: 79730474-79730474 on Build GRCh38
rs369392741 Chr.5: 79730549-79730549 on Build GRCh38
rs1032841211 Chr.5: 79730592-79730592 on Build GRCh38
rs773604802 Chr.5: 79730440-79730440 on Build GRCh38
rs764200838 Chr.5: 79730476-79730476 on Build GRCh38
rs747687553 Chr.5: 79730417-79730417 on Build GRCh38
rs766200568 Chr.5: 79730491-79730491 on Build GRCh38
rs944157774 Chr.5: 79730432-79730432 on Build GRCh38
rs778505660 Chr.5: 79730414-79730414 on Build GRCh38
rs564062961 Chr.5: 79730497-79730497 on Build GRCh38
rs746262176 Chr.5: 79730558-79730558 on Build GRCh38
rs16877124 Chr.5: 79730537-79730537 on Build GRCh38
rs530902441 Chr.5: 79730533-79730533 on Build GRCh38

View Full Product Details