Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34716266-34716539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC6
Entrez Gene ID
54102
Gene Name
chloride intracellular channel 6
Gene Aliases
CLIC1L
UniGene
Hs.473695
Gene Chromosome Location
Chr.21: 34668953-34718227 on Build GRCh38
Transcripts
NM_001317009.1 NM_053277.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4295980 c.2042A>C p.E681A Chr.21: 34716409-34716409 on Build GRCh38
7361530 c.1999A>G p.T667A Chr.21: 34716366-34716366 on Build GRCh38
5590236 c.1984G>A p.D662N Chr.21: 34716405-34716405 on Build GRCh38
5605640 c.1919G>A p.R640K Chr.21: 34716340-34716340 on Build GRCh38
6287637 c.2071G>C p.E691Q Chr.21: 34716438-34716438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372452162 Chr.21: 34716488-34716488 on Build GRCh38
rs757517803 Chr.21: 34716463-34716463 on Build GRCh38
rs770916000 Chr.21: 34716375-34716375 on Build GRCh38
rs143913704 Chr.21: 34716366-34716366 on Build GRCh38
rs375778168 Chr.21: 34716509-34716509 on Build GRCh38
rs756698489 Chr.21: 34716319-34716319 on Build GRCh38
rs374240622 Chr.21: 34716301-34716301 on Build GRCh38
rs772153839 Chr.21: 34716394-34716394 on Build GRCh38
rs115008561 Chr.21: 34716340-34716340 on Build GRCh38
rs761594203 Chr.21: 34716439-34716439 on Build GRCh38
rs772315358 Chr.21: 34716489-34716489 on Build GRCh38
rs141493110 Chr.21: 34716399-34716399 on Build GRCh38
rs767983391 Chr.21: 34716450-34716450 on Build GRCh38
rs763867886 Chr.21: 34716306-34716306 on Build GRCh38
rs145328525 Chr.21: 34716422-34716422 on Build GRCh38
rs199702073 Chr.21: 34716376-34716376 on Build GRCh38
rs773644106 Chr.21: 34716332-34716332 on Build GRCh38
rs775434573 Chr.21: 34716288-34716288 on Build GRCh38
rs558210239 Chr.21: 34716325-34716325 on Build GRCh38
rs763998020 Chr.21: 34716421-34716421 on Build GRCh38
rs377400896 Chr.21: 34716367-34716367 on Build GRCh38
rs183771003 Chr.21: 34716461-34716461 on Build GRCh38
rs778806319 Chr.21: 34716484-34716484 on Build GRCh38
rs756513353 Chr.21: 34716371-34716371 on Build GRCh38
rs1016134525 Chr.21: 34716447-34716447 on Build GRCh38
rs772395526 Chr.21: 34716297-34716297 on Build GRCh38
rs140169642 Chr.21: 34716449-34716449 on Build GRCh38
rs749972705 Chr.21: 34716446-34716446 on Build GRCh38
rs1007123477 Chr.21: 34716322-34716322 on Build GRCh38
rs776464087 Chr.21: 34716442-34716442 on Build GRCh38
rs546336753 Chr.21: 34716471-34716471 on Build GRCh38
rs781488184 Chr.21: 34716342-34716342 on Build GRCh38
rs775383196 Chr.21: 34716395-34716395 on Build GRCh38
rs1030089767 Chr.21: 34716379-34716379 on Build GRCh38
rs752005065 Chr.21: 34716337-34716337 on Build GRCh38
rs764584294 Chr.21: 34716444-34716444 on Build GRCh38
rs78180107 Chr.21: 34716372-34716372 on Build GRCh38
rs753564156 Chr.21: 34716313-34716313 on Build GRCh38
rs747000104 Chr.21: 34716508-34716508 on Build GRCh38
rs562673242 Chr.21: 34716455-34716455 on Build GRCh38
rs775542790 Chr.21: 34716499-34716499 on Build GRCh38
rs745738330 Chr.21: 34716382-34716382 on Build GRCh38
rs761184477 Chr.21: 34716350-34716350 on Build GRCh38
rs1043246904 Chr.21: 34716292-34716292 on Build GRCh38

View Full Product Details