Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10800928-10801153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G2
Entrez Gene ID
1982
Gene Name
eukaryotic translation initiation factor 4 gamma 2
Gene Aliases
AAG1, DAP5, NAT1, P97
UniGene
Hs.183684
Gene Chromosome Location
Chr.11: 10797046-10809035 on Build GRCh38
Transcripts
NM_001042559.2 NM_001172705.1 NM_001418.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652978 c.1529C>A p.P510H Chr.11: 10800972-10800972 on Build GRCh38
6343140 c.1539+9C>A p.? Chr.11: 10800953-10800953 on Build GRCh38
7366066 c.1488G>C p.M496I Chr.11: 10801013-10801013 on Build GRCh38
6131041 c.1501G>A p.A501T Chr.11: 10801000-10801000 on Build GRCh38
1351075 c.1536A>G p.G512G Chr.11: 10800965-10800965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024084788 Chr.11: 10801022-10801022 on Build GRCh38
rs916990902 Chr.11: 10801108-10801108 on Build GRCh38
rs749966549 Chr.11: 10801097-10801097 on Build GRCh38
rs143622080 Chr.11: 10801064-10801064 on Build GRCh38
rs754870131 Chr.11: 10801111-10801111 on Build GRCh38
rs778459575 Chr.11: 10800973-10800973 on Build GRCh38
rs11542382 Chr.11: 10801060-10801060 on Build GRCh38
rs372779966 Chr.11: 10801058-10801058 on Build GRCh38
rs35264886 Chr.11: 10801025-10801025 on Build GRCh38
rs187362968 Chr.11: 10801096-10801096 on Build GRCh38
rs746824736 Chr.11: 10800991-10800991 on Build GRCh38
rs750398533 Chr.11: 10800979-10800979 on Build GRCh38
rs748385557 Chr.11: 10801126-10801126 on Build GRCh38
rs780085043 Chr.11: 10800987-10800987 on Build GRCh38
rs113930127 Chr.11: 10800961-10800961 on Build GRCh38
rs1050514944 Chr.11: 10800984-10800984 on Build GRCh38
rs781607544 Chr.11: 10801004-10801004 on Build GRCh38
rs762974832 Chr.11: 10801046-10801046 on Build GRCh38
rs781121315 Chr.11: 10801116-10801116 on Build GRCh38
rs112725418 Chr.11: 10800960-10800960 on Build GRCh38
rs753445529 Chr.11: 10800958-10800958 on Build GRCh38
rs762874293 Chr.11: 10801042-10801042 on Build GRCh38
rs766452552 Chr.11: 10801101-10801101 on Build GRCh38
rs1054123676 Chr.11: 10800990-10800990 on Build GRCh38
rs769823449 Chr.11: 10801041-10801041 on Build GRCh38
rs763756463 Chr.11: 10800957-10800957 on Build GRCh38
rs959645819 Chr.11: 10801125-10801125 on Build GRCh38
rs145064111 Chr.11: 10800992-10800992 on Build GRCh38
rs761526926 Chr.11: 10801091-10801091 on Build GRCh38
rs1000521772 Chr.11: 10800983-10800983 on Build GRCh38
rs758364463 Chr.11: 10800985-10800985 on Build GRCh38
rs1049560512 Chr.11: 10800967-10800967 on Build GRCh38
rs1009976472 Chr.11: 10801008-10801008 on Build GRCh38
rs377419984 Chr.11: 10801107-10801107 on Build GRCh38
rs753533530 Chr.11: 10801082-10801082 on Build GRCh38
rs890568815 Chr.11: 10800989-10800989 on Build GRCh38
rs754266342 Chr.11: 10801102-10801102 on Build GRCh38
rs771480888 Chr.11: 10801045-10801045 on Build GRCh38
rs995107777 Chr.11: 10801087-10801087 on Build GRCh38
rs755326493 Chr.11: 10801125-10801126 on Build GRCh38

View Full Product Details