Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 681461-681688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL3
Entrez Gene ID
10272
Gene Name
follistatin like 3
Gene Aliases
FLRG, FSRP
UniGene
Hs.529038
Gene Chromosome Location
Chr.19: 676389-683392 on Build GRCh38
Transcripts
NM_005860.2

SNP Detail

SNP ID Chromosome Location
rs745996532 Chr.19: 681508-681508 on Build GRCh38
rs558755662 Chr.19: 681632-681632 on Build GRCh38
rs549983230 Chr.19: 681545-681545 on Build GRCh38
rs776547042 Chr.19: 681500-681500 on Build GRCh38
rs145117631 Chr.19: 681523-681523 on Build GRCh38
rs939405510 Chr.19: 681521-681521 on Build GRCh38
rs749972809 Chr.19: 681482-681482 on Build GRCh38
rs764621697 Chr.19: 681616-681616 on Build GRCh38
rs767250155 Chr.19: 681540-681540 on Build GRCh38
rs747419907 Chr.19: 681569-681569 on Build GRCh38
rs769857914 Chr.19: 681576-681576 on Build GRCh38
rs761509739 Chr.19: 681539-681539 on Build GRCh38
rs763425741 Chr.19: 681608-681608 on Build GRCh38
rs757386452 Chr.19: 681572-681572 on Build GRCh38
rs758932744 Chr.19: 681557-681557 on Build GRCh38
rs377720232 Chr.19: 681490-681490 on Build GRCh38
rs568510894 Chr.19: 681546-681546 on Build GRCh38
rs376159831 Chr.19: 681637-681637 on Build GRCh38
rs779514360 Chr.19: 681487-681487 on Build GRCh38
rs868447287 Chr.19: 681664-681664 on Build GRCh38
rs772909733 Chr.19: 681657-681657 on Build GRCh38
rs774321743 Chr.19: 681533-681533 on Build GRCh38
rs1030737059 Chr.19: 681651-681651 on Build GRCh38
rs772863832 Chr.19: 681598-681598 on Build GRCh38
rs368764119 Chr.19: 681583-681583 on Build GRCh38
rs769266104 Chr.19: 681484-681486 on Build GRCh38
rs749869791 Chr.19: 681544-681544 on Build GRCh38
rs781540655 Chr.19: 681575-681575 on Build GRCh38
rs372400894 Chr.19: 681593-681593 on Build GRCh38
rs376133733 Chr.19: 681526-681526 on Build GRCh38
rs748048439 Chr.19: 681653-681653 on Build GRCh38
rs900931778 Chr.19: 681617-681617 on Build GRCh38
rs753365926 Chr.19: 681603-681603 on Build GRCh38
rs775559948 Chr.19: 681518-681518 on Build GRCh38
rs758184758 Chr.19: 681588-681588 on Build GRCh38
rs201785071 Chr.19: 681563-681563 on Build GRCh38
rs147593429 Chr.19: 681655-681655 on Build GRCh38
rs763917132 Chr.19: 681532-681532 on Build GRCh38
rs768737961 Chr.19: 681585-681585 on Build GRCh38
rs751913369 Chr.19: 681619-681619 on Build GRCh38
rs200494419 Chr.19: 681600-681600 on Build GRCh38
rs960371196 Chr.19: 681665-681665 on Build GRCh38
rs750095878 Chr.19: 681484-681484 on Build GRCh38
rs928073841 Chr.19: 681501-681501 on Build GRCh38
rs780289220 Chr.19: 681580-681580 on Build GRCh38
rs534075695 Chr.19: 681621-681621 on Build GRCh38
rs199545804 Chr.19: 681489-681489 on Build GRCh38
rs760248530 Chr.19: 681599-681599 on Build GRCh38
rs984975120 Chr.19: 681494-681494 on Build GRCh38
rs779060887 Chr.19: 681644-681644 on Build GRCh38
rs769618663 Chr.19: 681511-681511 on Build GRCh38
rs772562730 Chr.19: 681595-681595 on Build GRCh38
rs753134505 Chr.19: 681547-681547 on Build GRCh38
rs144316081 Chr.19: 681496-681496 on Build GRCh38
rs780234224 Chr.19: 681639-681639 on Build GRCh38
rs749429626 Chr.19: 681643-681643 on Build GRCh38
rs923286991 Chr.19: 681536-681536 on Build GRCh38
rs1039741665 Chr.19: 681602-681602 on Build GRCh38

View Full Product Details